Skip to Content
Merck
All Photos(1)

Key Documents

ABC846

Sigma-Aldrich

Anti-Slc22A17 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Solute carrier family 22 member 17, 24p3 receptor, 24p3R, Brain-type organic cation transporter, Lipocalin-2 receptor, Neutrophil gelatinase-associated lipocalin receptor, NgalR, Slc22A17

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€370.00

€370.00


Estimated to ship on25 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
€370.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€370.00


Estimated to ship on25 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

immunofluorescence: suitable
western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

General description

Solute carrier family 22 member 17, also known as 24p3 receptor (24p3R) or Brain-type organic cation transporter or Lipocalin-2 receptor or Neutrophil gelatinase-associated lipocalin receptor (NgalR), and encoded by the gene name SLC22A17 or BOCT or BOIT, is a family of organic anion and cation transporters (OATs, OCTs, OCTNs) that are expressed in brain. Each contain 12 predicted alpha-helical transmembrane domains (TMDs) and one large extracellular loop between TMDs 1 and 2. Transporters of the SLC22 family function in different ways such as uniporters that mediate facilitated diffusion in either direction (OCTs), as anion exchangers (OAT1, OAT3 and URAT1), and as Na(+)/l-carnitine cotransporter (OCTN2). Slc22 family members participate in the absorption and/or excretion of drugs, xenobiotics, and endogenous compounds in intestine, liver, and kidney, and perform homeostatic functions in brain and heart. Mutations in the Slc22 family may cause specific diseases such as primary systemic carnitine deficiency or idiopathic renal hypouricemia and may change drug absorption or excretion. Recent studies show the expression of Slc22A17 as receptor for Lipocalin 2 is relatively high in hematopoietic stem cells.

Immunogen

KLH-conjugated synthetic peptide corresponding to the C-terminus of human Slc22A17.

Application

Anti-Slc22A17 Antibody is an antibody against Slc22A17 for use in western blotting & immunofluorescence.
Immunofluorescence Analysis: 20 μg/mL from a representative lot detected Slc22A17 in rat kidney tissue.

Quality

Evaluated by Western Blotting in SK-N-SH cell lysate with and without blocking peptide.

Western Blotting Analysis: 1 μg/mL of this antibody detected Slc22A17 in SK-N-SH cell lysate with and without blocking peptide.

Target description

~60 kDa observed. Uncharacterized bands may be observed in some lysate(s).

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service