Skip to Content
Merck
All Photos(2)

Key Documents

ABC841

Sigma-Aldrich

Anti-Caspase-14 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Caspase-14, CASP-14

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€446.00

€446.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
€446.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€446.00


Estimated to ship on28 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

immunohistochemistry: suitable
western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CASP14(23581)

General description

Caspases are a family of cysteine proteases that can be divided into apoptotic and inflammatory caspase subfamilies. Caspase-14 exists as 2 subunits (p19 and p10) and is highly expressed in embryonic but not adult tissues. It is processed and activated by caspase 8 and caspase 10 in vitro, and by anti-Fas agonist antibody or TNF-related apoptosis inducing ligand in vivo. The expression and processing of this caspase may be involved in the keratinocyte terminal differentiation, which is important for the formation of the skin barrier.

Specificity

This antibody detects the pro-form of Caspase-14 as well as the large subunit cleaved form.

Immunogen

KLH-conjugated linear peptide corresponding to 16 amino acids near the C-terminus of human Caspase-14.

Application

Anti-Caspase-14 Antibody is a highly specific rabbit polyclonal antibody, that targets Caspase 14 & has been tested in western blotting & IHC.
Immunohistochemistry Analysis: 2.5 μg/mL from a representative lot detected Caspase-14 in human liver tissue.

Quality

Evaluated by Western Blotting in Jurkat cell lysate with and without blocking peptide.

Western Blotting Analysis: 1 μg/mL of this antibody detected Caspase-14 in Jurkat cell lysate with and without blocking peptide.

Target description

~28 kDa observed. Uncharacterized bands may be observed in some lysate(s).

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service