Skip to Content
Merck

ABC477

Anti-YY1AP1 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

YY1-associated protein 1, Hepatocellular carcinoma susceptibility protein, Hepatocellular carcinoma-associated protein 2

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IHC, WB
Citations:
1
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, human, rat

species reactivity (predicted by homology)

chimpanzee (based on 100% sequence homology)

technique(s)

immunohistochemistry: suitable, western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... YY1AP1(55249)

General description

YY1-associated protein 1 (YY1AP1), also known as Hepatocellular carcinoma susceptibility protein, Hepatocellular carcinoma-associated protein 2, is encoded by the gene name YY1AP1, HCCA2, and YY1AP. YY1AP1 has been demonstrated to enhance transcription activation by interacting with YY1. Recent studies have shown that YY1AP1 may play a role in cell cycle regulation. YY1AP1 interacts with YY1 and MAD2L2. YY1AP1 is expressed ubiquitously. YY1AP1 has been detected in small intestine, skeletal muscle, lung, pancreas, brain, stomach, spleen, colon and heart. At very low levels, YY1AP1 has been detected in healthy liver. YY1AP1 is highly expressed in most liver carcinomas.
~90 kDa observed. The endogenous YY1AP protein signal is found at 90 kDa, in agreement with the predicted protein size of 750 amino acids derived from open reading frame (Wang, C.Y., et al. (2004). J. Biol. Chem. 279:17750-17755).

Immunogen

Epitope: Near N-terminal
KLH-conjugated linear peptide corresponding to human YY1AP1 near the N-terminal.

Application

Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional
Western Blotting Analysis: 0.5 µg/mL from a representative lot detected YY1AP1 in 10 µg of C2C12, Hek293, HeLa, HepG2, human placenta, Huvec, Jurkat, L6, mouse brain, NIH-3T3, PC12, PC3, and Raw 264.7 lysate.
Immunohistochemistry Analysis: A 1:50-250 dilution from a representative lot detected YY1AP1 in human cerebellum, mouse hindbrain, and mouse cerebellum tissue.
This Anti-YY1AP1 antibody is validated for use in WB, IH for the detection of YY1AP1.

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Evaluated by Western Blotting in A431 cell lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected YY1AP1 in 10 µg of A431 cell lysate.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library







Global Trade Item Number

SKUGTIN
ABC47704055977122152