Skip to Content
Merck

Skip To

ABC42-M

Anti-Mitofusin-2 (Mfn2) Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Mitofusin-2, Transmembrane GTPase MFN2

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 μg
Check Cart for Availability
€ 401,00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
WB
Citations:
1

€ 401,00


Check Cart for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat, human

species reactivity (predicted by homology)

mouse (based on 100% sequence homology)

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... MFN2(9927)

General description

Mitofusin-2 (Mfn2) is a membrane protein of the mitochondria and is important in mitochondrial fusion in mammalian cells. Mfn2 activity has also been implicated in mitochondrial metabolism and the loss of Mfn2 may be the reason for certain metabolic modifications that are seen in individuals with obesity. Mfn2 has been associated with maintaining the membrane potential of the mitochondria. Mfn2 has been shown to hinder mitochondrial antiviral signaling by inhibiting production of type 1 interferons. Mfn2 is a p53-inducible target and involved in cell proliferation, apoptosis promotion and tumor suppression.
~86 kDa observed.
Mitofusion-2 (Mfn2) exists as 2 isoforms produced by alternative splicing with molecular weights of ~86 kDa and ~50 kDa

Immunogen

Epitope: Coiled Coil Region of the Cytoplasmic Domain
KLH-conjugated linear peptide corresponding to the cytoplasmic domain of human Mitofusin-2 (Mfn2).

Application

Detect Mitofusin-2 (Mfn2) using this Anti-Mitofusin-2 (Mfn2) Antibody validated for use in WB.

Biochem/physiol Actions

This antibody recognizes Mitofusin-2 (Mfn2) at the coiled coil region of the cytoplasmic domain.

Analysis Note

Evaluated by Western Blot in human heart tissue lysate.

Western Blot Analysis: 0.5 µg/mL of this antibody detected Mitofusion-2 (Mfn2) in 10 µg of human heart tissue lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
ABC41-MM6319WH0009927M3
Quality Level

100

Quality Level

100

Quality Level

200

Quality Level

100

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

biological source

rabbit

biological source

rabbit

biological source

rabbit

biological source

mouse

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

purified immunoglobulin

technique(s)

western blot: suitable

technique(s)

western blot: suitable

technique(s)

immunoprecipitation (IP): 5-10 μg using HeLa human epitheloid carcinoma cell lysate, indirect immunofluorescence: 20-30 μg/mL using differentiated mouse C2 cells, western blot (chemiluminescent): 0.5-1 μg/mL using extracts of rat or mouse brain mitochondria

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable, indirect ELISA: suitable, western blot: 1-5 μg/mL

UniProt accession no.

O95140

UniProt accession no.

Q8IWA4

UniProt accession no.

O95140

UniProt accession no.

O95140


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters