Skip to Content
Merck

ABC338

Anti-IRGM Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

IRGM Antibody, Immunity related GTPase family, IFL1, IRGM1, LRG47, LRG-47

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IF, IHC, WB
Citations:
1
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

immunofluorescence: suitable, immunohistochemistry: suitable (paraffin), western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... IRGM(345611)

General description

Immunity-related GTPase family M protein (IRGM), also known as Interferon-inducible protein 1 (IFI1) and LPS-stimulated RAW 264.7 macrophage protein 47 homolog (LRG-47), is a member of the interferon-inducible GTPase family. It induces autophagy and generates large autolysosomal organelles as a mechanism for the elimination of intracellular Mycobacterium tuberculosis. IRGM is expressed in colon, small bowel, and peripheral blood leukocytes. Mutations in IRGM have been assoicated with inflammatory bowel disease type 19 which is subdivided into Crohn′s disease and ulcerative colitis phenotypes.
~31 kDa observed. Uncharacterized bands may appear in some lysates.

Immunogen

KLH-conjugated linear peptide corresponding to the intermediate domain of human IRGM.

Application

Detect Immunity-related GTPase family M protein using this rabbit polyclonal antibody, Anti-IRGM Antibody validated for use in western blotting, IHC (Paraffin) & Immunofluorescence.
Immunohistochemistry Analysis: 5 µg/mL from a representative lot detected IRGM in human brain tissue.

Immunofluorescence Analysis: 20 μg/mL from a representative lot detected IRGM in human brain cells.

Biochem/physiol Actions

This antibody recognizes the intermediate domain of IRGM.

Analysis Note

Evaluated by Western Blotting in human brain tissue lysate.

Western Blotting Analysis: 2 µg/mL of this antibody detected IRGM in 15 µg of human brain tissue lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library







Global Trade Item Number

SKUGTIN
ABC33804053252879791