Skip to Content
Merck

Skip To

ABC334

Anti-MAP1 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Modulator of apoptosis 1, MAP-1, MAP1, Paraneoplastic antigen Ma4

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 μg
Check Cart for Availability
€ 340,00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
WB
Citations:
-

€ 340,00


Check Cart for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse

species reactivity (predicted by homology)

human (immunogen homology)

technique(s)

western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... MOAP1(64112)

General description

Modulator of apoptosis 1 (MAP1) is a member of the PNMA family. This protein contains a BH3-like domain and induces caspase-dependent apoptosis in mammalian cells when overexpressed and is required for death receptor-dependent apoptosis. MAP1 is ubiquitously expressed with higher levels found in heart and brain.
~35 kDa observed

Immunogen

KLH-conjugated linear peptide corresponding to the intermediate domain of human MAP1.

Application

Detect Modulator of apoptosis 1 using this rabbit polyclonal antibody, Anti-MAP1 Antibody validated for use in western blotting.

Biochem/physiol Actions

This antibody recognizes the intermediate domain of MAP1.

Analysis Note

Evaluated by Western Blotting in EL4 cell lysate.

Western Blotting Analysis: 1 µg/mL of this antibody detected MAP1 in 15 µg of EL4 cell lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SAB1407857M4528HPA076948
biological source

rabbit

biological source

mouse

biological source

mouse

biological source

rabbit

Quality Level

100

Quality Level

100

Quality Level

200

Quality Level

100

antibody form

affinity isolated antibody

antibody form

purified immunoglobulin

antibody form

ascites fluid

antibody form

affinity isolated antibody

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

species reactivity

mouse

species reactivity

human

species reactivity

human, feline, bovine, hamster, chicken, mouse, rat

species reactivity

human

technique(s)

western blot: suitable

technique(s)

western blot: 1 μg/mL

technique(s)

microarray: suitable, western blot: 1:500 using a fresh total rat brain extract or an enriched microtubule protein preparation

technique(s)

immunofluorescence: 0.25-2 μg/mL (ICC-IF), immunohistochemistry: 1:500 - 1:1000


Still not finding the right product?

Try our Product Selector Tool to narrow your options


wgk

WGK 2

Storage Class

12 - Non Combustible Liquids



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters