Sign In to View Organizational & Contract Pricing.
About This Item
UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IHC, WB
Citations:
-
biological source
rabbit
Quality Level
conjugate
unconjugated
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
purified by
affinity chromatography
species reactivity
human
technique(s)
immunohistochemistry: suitable, western blot: suitable
UniProt accession no.
General description
Partner and localizer of BRCA2 (PALB2) is the critical protein required for the homologous recombination repair at DNA breaks. It serves as a scaffold protein in the formation of the BRAC1-PALB2-BRAC2 complex which is essential for homologous recombination. Essentially a partner of BRAC2, PALB2 localizes BRAC2 and stabilizes it. PALB2 enables BRAC2 to perform its recombinational repair and checkpoint functions in addition to its DNA repair functions. Also by binding BRAC2, PALB2 is thought to help BRCA2 avoid degradation. PALB2 localizes to the nucleus and has at least four known isoforms. Defects in PALB2 are linked with many forms of breast cancers, pancreatic cancer type 3 (PNCA3), as well as disorders of the bone marrow (Fanconi anemia complementation group, FANCN). These patients suffer numerous cardiac, limb, and skin malformations as well as malignancies. EMD Millipore’s Anti-PALB2 rabbit polyclonal antibody has been tested in western blotting in human SK-N-SH cell lysate samples.
~180 kDa observed. Uncharacterized bands may appear in some lysate(s).
Immunogen
KLH-conjugated linear peptide corresponding to human PALB2.
Application
Immunohistochemistry Analysis: A 1:250-1,000 dilution from a representative lot detected SPALB2 in human breast cancer tissue.
Optimal working dilutions must be determined by end user.
Optimal working dilutions must be determined by end user.
Anti-PALB2 Antibody is a rabbit polyclonal antibody, validated for use in western blotting & IHC.
Analysis Note
Evaluated by Western Blotting in SK-N-SH cell lysate.
Western Blotting Analysis: 2 µg/mL of this antibody detected PALB2 in 15 µg of SK-N-SH cell lysate.
Western Blotting Analysis: 2 µg/mL of this antibody detected PALB2 in 15 µg of SK-N-SH cell lysate.
Other Notes
Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.
Not finding the right product?
Try our Product Selector Tool.
Storage Class
12 - Non Combustible Liquids
wgk
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
Global Trade Item Number
| SKU | GTIN |
|---|---|
| ABC331 | 04053252917387 |
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service