Skip to Content
Merck

Skip To

ABC2075

Anti-LC3C

from rabbit, purified by affinity chromatography

Synonym(s):

Microtubule-associated proteins 1A/1B light chain 3C, Autophagy-related protein LC3 C, Autophagy-related ubiquitin-like modifier LC3C, MAP1 light chain 3-like protein 3, MAP1A/MAP1B light chain 3 C, MAP1A/MAP1B LC3 C, Microtubule-associated protein 1 lig

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
50 μL
Check Cart for Availability
€ 134,00
200 μL
Check Cart for Availability
€ 447,00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IHC, WB
Citations:
-

€ 134,00


Check Cart for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat

species reactivity (predicted by homology)

human (based on 100% sequence homology)

technique(s)

immunohistochemistry: suitable (paraffin), western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... MAP1LC3C(440738)

General description

Microtubule-associated proteins 1A/1B light chain 3C (UniProt: Q9BXW4; also known as Autophagy-related protein LC3C, Autophagy-related ubiquitin-like modifier LC3C, MAP1 light chain 3-like protein 3, MAP1A/MAP1B light chain 3 C, MAP1A/MAP1B LC3 C, Microtubule-associated protein 1 light chain 3 gamma) is encoded by the MAP1LC3C gene (Gene ID: 440738) in human. LC3s (MAP1-LC3A, B and C) are structural proteins of autophagosomal membranes, widely used as biomarkers of autophagy. LC3A is mainly localized to perinuclear and nuclear regions, LC3B is evenly distributed throughout the cytoplasm and is also localized in the nucleolar regions, and LC3C is shown to be located in the cytoplasm and strongly in the nuclei. LC3C is synthesized with a propeptide (aa 127-147), which is removed in the mature form. LC3C is a ubiquitin-like modifier that plays a crucial role in antibacterial autophagy through the selective binding of CALCOCO2. It recruits all ATG8 family members to infecting bacteria such as S.Typhimurium. It is also reported to play a role in aggrephagy, the macroautophagic degradation of ubiquitinated and aggregated proteins. LC3C is recruited to damaged mitochondria where it mediates autophagosomal degradation of mitochondrial parts in a piecemeal fashion and is required for maintaining basal mitochondrial network homeostasis. It also mediates autophagosomal degradation of MTX1 protein and cooperates with p62 to deliver MTX1 to lysosomes. (Ref.: Koukourakis, KI et al. (2015). PLoS One DOI:10.1371/journal.pone.0137675; Le Guerroue´ et al., 2017, Mol. Cell 68(4); 786-796).
~17 kDa observed; 16.85 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Immunogen

Epitope: unknown
GST-tagged full-length human recombinant Microtubule-associated proteins 1A/1B light chain 3C (LC3C).

Application

Anti-LC3C, Cat. No. ABC2075, is a rabbit polyclonal antibody that detects Microtubule-associated proteins 1A/1B light chain 3C and has been tested for use in Immunohistochemistry (Paraffin) and Western Blotting.
Immunohistochemistry (Paraffin) Analysis: A 1:250 dilution from a representative lot detected LC3C in rat brain tissue sections.
Research Category
Apoptosis & Cancer

Biochem/physiol Actions

This rabbit polyclonal antibody specifically detects LC3C.

Physical form

Affinity Purified
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Evaluated by Western Blotting in rat brain tissue lysate.

Western Blotting Analysis: A 1:500 dilution of this antibody detected LC3C in rat brain tissue lysate.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SAB4200840ABC929ABC232
biological source

rabbit

biological source

mouse

biological source

rabbit

biological source

rabbit

conjugate

unconjugated

conjugate

unconjugated

conjugate

-

conjugate

-

antibody form

affinity isolated antibody

antibody form

-

antibody form

affinity isolated antibody

antibody form

serum

Gene Information

human ... MAP1LC3C(440738)

Gene Information

human ... MAP1LC3C(440738)

Gene Information

human ... MAP1A(4130), MAP1LC3A(84557)

Gene Information

human ... MAP1LC3A(84557)

UniProt accession no.

Q9BXW4

UniProt accession no.

Q9BXW4

UniProt accession no.

Q9H492

UniProt accession no.

Q9H492

technique(s)

immunohistochemistry: suitable (paraffin), western blot: suitable

technique(s)

immunoblotting: 0.5-1 μg/mL using whole extracts of HEK-293T cells overexpressing human LC3C

technique(s)

immunocytochemistry: suitable, western blot: suitable

technique(s)

western blot: suitable


Still not finding the right product?

Try our Product Selector Tool to narrow your options




Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters