Skip to Content
Merck
All Photos(2)

Key Documents

ABC152

Sigma-Aldrich

Anti-Annexin A2 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Annexin A2, Annexin II, Annexin-2, Calpactin I heavy chain, Calpactin-1 heavy chain, Chromobindin-8, Lipocortin II, Placental anticoagulant protein IV, PAP-IV, Protein I, p36

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat, human

species reactivity (predicted by homology)

primate (based on 100% sequence homology), mouse (based on 100% sequence homology), bovine (based on 100% sequence homology), porcine (based on 100% sequence homology), canine (based on 100% sequence homology)

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ANXA2(302)

General description

Annexin A2 is known by several other names including: Annexin II, Calpactin I heavy chain, Chromobindin-8, Lipocortin II and Placental anticoagulant protein IV. Annexin A2 is a calcium-regulated membrane-binding protein that binds two calcium ions with high affinity. Annexin A2′s affinity for calcium is enhanced by anionic phospholipids. Annexin A2 has been shown to bind to human cytomegalvirus (HCMV) and may be involved in heat-stress response and apoptosis.

Specificity

This antibody recognizes the Annexin 1 repeat domain of Annexin A2.

Immunogen

Epitope: Annexin 1 repeat domain
KLH-conjugated linear peptide corresponding to Annexin 1 repeat domain of human Annexin A2.

Application

Anti-Annexin A2 is an antibody targeting the Annexin A2 protein, validated for use in WB & IHC.
Immunohistochemistry Analysis: A 1:500 dilution from a representative lot detected Annexin A2 in human and rat colon tissues.
Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional

Quality

Evaluated by Western Blotting in A549 cell lysate.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected Annexin A2 in 10 µg of A549 cell lysate.

Target description

~36 kDa observed

Physical form

Protein A purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
A549 cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shahzadi Noreen et al.
Proteomics. Clinical applications, 14(2), e1900078-e1900078 (2019-11-21)
An early and accurate diagnosis of ovarian carcinoma (OC) may reduce morbidity and mortality of the patients. To improve the clinical outcome in OC patients, the present study is aimed at identifying robust biomarkers for early OC diagnosis. EXPERIMENTAL DESIGN: In

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service