Skip to Content
Merck

Skip To

ABC1466

Anti-phospho DCC (Tyr1418)

serum, from rabbit

Synonym(s):

Netrin receptor DCC, Tumor suppressor protein DCC, Deleted in Colorectal Cancer

Sign In to View Organizational & Contract Pricing.

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Clone:
polyclonal
Species reactivity:
rat
Application:
DB, IF, WB
Citations:
-


biological source

rabbit

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

rat

packaging

antibody small pack of 25 μL

technique(s)

dot blot: suitable, immunofluorescence: suitable, western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

target post-translational modification

phosphorylation (pTyr1418)

Gene Information

rat ... Dcc(25311)

General description

Netrin receptor DCC (UniProt: Q63155; also known as Tumor suppressor protein DCC, Deleted in Colorectal Cancer) is encoded by the Dcc gene (Gene ID: 25311) in rat. Netrin receptor DCC is a single-pass type I membrane protein that serves as a receptor for netrin and is essential for axon guidance. In response to netrin-1 stimulation, DCC becomes a signaling platform to recruit proteins that promote axon outgrowth and guidance. It is detected in embryonic spinal cord, predominantly in axons of commissural neurons. Deficiencies in netrin-1 or DCC expression can result in the loss of cerebral and spinal commissures. Netrin receptor DCC also acts as a dependence receptor required for apoptosis induction when not associated with netrin ligand and has been implicated as a tumor suppressor gene. It is synthesized with a signal peptide (aa 1-25), which is subsequently cleaved off. It has four Ig-like C2-type domains and six fibronectin type-III domains. In response to netrin-1 stimulation DCC is phosphorylated on serine, threonine, and tyrosine residues. Its phosphorylation on Tyrosine 1418 by Src family kinases is shown to be essential for netrin-1 to mediate axon outgrowth and guidance. It has been reported that in response to netrin-1, p120RasGAP is recruited to DCC in growth cones and forms a multiprotein complex with focal adhesion kinase (FAK) and extracellular signal regulated kinase (ERK). Activities of both Ras and ERK as shown to be elevated in p120RasGAP deficient neurons. (Ref.: Antoine-Bertrand J., et al., (2016). J. Biol. Chem. 291(9):4589-602).
~190 kDa observed; 157.94 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Immunogen

A linear peptide corresponding to 15 amino acids surrounding phospho-tyrosine 1418 from the C-terminal region of rat Netrin receptor DCC.
Epitope: C-terminus

Application

Anti-phospho DCC (Tyr1418), Cat. No. ABC1466, iis a rabit polyclonal antibody that detects Netrin receptor DCC and has been tested for use in Dot Blot, Immunofluorescence, and Western Blotting.
Dot Blot Analysis: A representative lot detected phospho DCC (Tyr1418) in Dot Blot applications (Antoine-Bertrand, J., et. al. (2016). J Biol Chem. 291(9):4589-602).

Immunofluorescence Analysis: A representative lot detected phospho DCC (Tyr1418) in Immunofluorescence applications (Plooster, M., et. al. (2017). Mol Biol Cell. 28(18):2374-2385).

Western Blotting Analysis: A representative lot detected phospho DCC (Tyr1418) in Western Blotting applications (Plooster, M., et. al. (2017). Mol Biol Cell. 28(18):2374-2385; Petrie, R.J., et. al. (2009). Biol Cell. 101(2):77-90).
Research Category
Apoptosis & Cancer

Biochem/physiol Actions

This rabbit polyclonal antibody detects Netrin receptor DCC phosphorylalted on Tyrosine 1418 in rat cells. It targets an epitope within 15 amino acids from the C-terminal region.

Physical form

Rabbit polyclonal antiserum with 0.05% sodium azide with 20% glycerol.
Unpurified

Preparation Note

Stable for 1 year at -20°C from date of receipt.

Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Evaluated by Western Blotting in HEK293 transfected with pRK5-DCC (DCCWT).

Western Blotting Analysis: A 1:500 dilution of this antibody detected phospho DCC (Tyr1418) in HEK293 transfected wtih pRK5-DCC (DCCWT).

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
ABE1950WH0004163M1MABN2292
antibody form

serum

antibody form

purified antibody

antibody form

purified immunoglobulin

antibody form

purified antibody

biological source

rabbit

biological source

rabbit

biological source

mouse

biological source

rat

species reactivity

rat

species reactivity

human

species reactivity

human

species reactivity

human

clone

polyclonal

clone

polyclonal

clone

1A7, monoclonal

clone

4F10, monoclonal

Gene Information

rat ... Dcc(25311)

Gene Information

human ... CCAR2(57805)

Gene Information

human ... MCC(4163)

Gene Information

human ... NRG1(3084)

technique(s)

dot blot: suitable, western blot: suitable, immunofluorescence: suitable

technique(s)

ChIP: suitable, immunocytochemistry: suitable, immunohistochemistry: suitable, immunoprecipitation (IP): suitable, western blot: suitable

technique(s)

indirect ELISA: suitable, western blot: 1-5 μg/mL

technique(s)

western blot: suitable


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters