Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|---|---|
100 μL | Check Cart for Availability | € 467,00 |
About This Item
UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
WB
Citations:
2
biological source
rabbit
Quality Segment
conjugate
unconjugated
antibody form
serum
antibody product type
primary antibodies
clone
polyclonal
species reactivity
mouse, rat, human
technique(s)
western blot: suitable
NCBI accession no.
UniProt accession no.
shipped in
wet ice
target post-translational modification
unmodified
Gene Information
human ... PEX13(5194)
General description
Peroxisomal membrane protein 13 (PEX13) is a component of the peroxisomal translocation machinery, which binds to peroxisomal targeting signal receptors via its SH3 domain, and facilitates the import of these receptors into the peroxisome membrane. Defects in PEX13 have been linked to neurodegenerative disorders, such as Zellweger syndrome.
~49 kDa observed. This protein has an expected molecular weight of 44 kDa; however, it has been observed at ~49 kDa (Muller, C. C., et al. 2011. Dis Model Mech. 4(1):104-110.). An uncharacterized band may be observed at ~41 kDa in some cell lysates.
Immunogen
Recombinant protein corresponding to human PEX13 (peroxisomal membrane marker 13).
Application
Research Category
Inflammation & Immunology
Inflammation & Immunology
Research Sub Category
Inflammation & Autoimmune Mechanisms
Inflammation & Autoimmune Mechanisms
This Anti-PEX13 (peroxisomal membrane marker) Antibody is validated for use in Western Blotting, ICC for the detection of PEX13 (peroxisomal membrane marker).
Physical form
Rabbit polyclonal serum with 0.05% sodium azide.
Unpurified
Preparation Note
Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Analysis Note
Control
HepG2 cell lysate
HepG2 cell lysate
Evaluated by Western Blot in HepG2 cell lysate.
Western Blot Analysis: A 1:1,000 dilution of this antibody detected PEX13 (peroxisomal membrane marker) in HepG2 cell lysate.
Western Blot Analysis: A 1:1,000 dilution of this antibody detected PEX13 (peroxisomal membrane marker) in HepG2 cell lysate.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
1 of 1
This Item | |||
|---|---|---|---|
| antibody form serum | antibody form serum | antibody form purified immunoglobulin | antibody form purified immunoglobulin |
| biological source rabbit | biological source rabbit | biological source mouse | biological source mouse |
| conjugate unconjugated | conjugate unconjugated | conjugate unconjugated | conjugate unconjugated |
| Quality Level 100 | Quality Level 100 | Quality Level 100 | Quality Level - |
| Gene Information human ... PEX13(5194) | Gene Information human ... PEX14(5195) | Gene Information human ... PEX14(5195) | Gene Information human ... PEX19(5824) |
| UniProt accession no. | UniProt accession no. | UniProt accession no. | UniProt accession no. |
Still not finding the right product?
Try our Product Selector Tool to narrow your options
Storage Class
10 - Combustible liquids
wgk
WGK 1
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



