Skip to Content
Merck
All Photos(3)

Key Documents

ABC137

Sigma-Aldrich

Anti-ATPIF1 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

ATPase inhibitor, mitochondrial, Inhibitor of F(1)F(o)-ATPase, IF(1), IF1, ATPIF1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€475.00

€475.00


Estimated to ship on21 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
€475.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€475.00


Estimated to ship on21 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, mouse

technique(s)

immunocytochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ATPIF1(93974)

General description

ATPIF1 is an interesting protein that acts on ATP synthase and prevents it from catabolizing ATP when the mitochondrial membrane potential falls below a threshold, which ATP synthase starts to do by hydrolyzing ATP to pump protons out of the mitochondrial matrix to restore ion balance. ATPIF1 also indirectly regulates heme synthesis by modulating the mitochondrial pH and redox potential. ATPIF1 is expressed in the mitochondria and its active is determined by conformation. Homodimers are active, homotetramers are inactive and their formation is governed by pH. ATPIF1, because of its effects on ATP and heme, is important in cellular development, heme biosynthesis, energy metabolism and angiogenesis and erythrocytes differentiation.

Immunogen

Epitope: N-terminal inhibitory region & Antiparallel alpha-helical coiled region
GST-tagged recombinant protein corresponding to the N-terminal inhibitory region & Antiparallel alpha-helical coiled eegion of human ATPIF1.

Application

Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional
This Anti-ATPIF1 Antibody is validated for use in Western Blotting and Immunocytochemistry for the detection of ATPIF1.
Western Blotting Analysis: 2.0 µg/mL from a representative lot detected ATPIF1 in 10 µg of HEK293 cell lysate.
Immunocytochemistry Analysis: A 1:500 dilution from a representative lot detected ATPIF1 in HeLa, A431, and NIH/3T3 cells.

Quality

Evaluated by Western Blotting in T47D cell lysate.

Western Blotting Analysis: 2.0 µg/mL of this antibody detected ATPIF1 in 10 µg of T47D cell lysate.

Target description

~12 kDa observed. Uncharacterized band(s) may appear in some lysates.

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Octavian Bucur et al.
Nature protocols, 15(5), 1649-1672 (2020-04-03)
In pathology, microscopy is an important tool for the analysis of human tissues, both for the scientific study of disease states and for diagnosis. However, the microscopes commonly used in pathology are limited in resolution by diffraction. Recently, we discovered

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service