Sign In to View Organizational & Contract Pricing.
About This Item
UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
ICC, IHC, IP, WB
Citations:
3
biological source
rabbit
Quality Segment
conjugate
unconjugated
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
purified by
affinity chromatography
species reactivity
mouse, rat, human
technique(s)
immunocytochemistry: suitable, immunohistochemistry: suitable, immunoprecipitation (IP): suitable, western blot: suitable
NCBI accession no.
UniProt accession no.
shipped in
wet ice
target post-translational modification
unmodified
Gene Information
human ... BAK1(578)
General description
Bak or Bcl2 homologous antagonist is a member of the Bcl2 family of proteins. The Bcl-2 related proteins interact with one another through the formation of homo and heterodimers. The susceptibility of cells to apoptotic stimuli is thought to be controlled by the relative ratios of the different Bcl2 family proteins. Bak has been demonstrated to accelerate the rate of apoptosis in growth factor deprived murine lymphoid, neuronal and fibroblastic cell lines. This protein localizes to mitochondria, and functions to induce apoptosis. It interacts with and accelerates the opening of the mitochondrial voltage-dependent anion channel, which leads to a loss in membrane potential and the release of cytochrome c. Bak also interacts with the tumor suppressor P53 after exposure to cell stress.
~ 24 kDa
Immunogen
KLH-conjugated linear peptide corresponding to human BAK at and around the N-terminus.
Application
Detect Bak (N-terminus) using this Anti-Bak (N-terminus) Antibody validated for use in WB, IH, IC & IP.
Western Blot (SNAP ID) Analysis: 1 - 4 µg/ml from a previous lot detected Bak on 10 µg of Hek293 cell lysate.
Immunoprecipitation Analysis: For unpurified antibodies. 10 µg from a previous lot immunoprecipitated Bak from 500 µg of NIH3T3 lysate.
Immunohistochemistry Analysis: 1:300 dilution from a previous lot detected Bak in kidney tissue.
Immunocytochemistry Analysis: 1:500 dilution from a previous lot detected Bak in HeLa, NIH/3T3, and A431 cells.
Immunoprecipitation Analysis: For unpurified antibodies. 10 µg from a previous lot immunoprecipitated Bak from 500 µg of NIH3T3 lysate.
Immunohistochemistry Analysis: 1:300 dilution from a previous lot detected Bak in kidney tissue.
Immunocytochemistry Analysis: 1:500 dilution from a previous lot detected Bak in HeLa, NIH/3T3, and A431 cells.
Biochem/physiol Actions
This antibody recognizes BAK.
Analysis Note
Evaluated by Western Blot in Hek293 cell lysate.
Western Blot Analysis: 0.5 µg/ml of this antibody detected Bak on 10 µg of Hek293 cell lysate.
Western Blot Analysis: 0.5 µg/ml of this antibody detected Bak on 10 µg of Hek293 cell lysate.
Other Notes
Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.
Replaces: 04-433
Still not finding the right product?
Try our Product Selector Tool to narrow your options
Storage Class
12 - Non Combustible Liquids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.