Skip to Content
Merck
All Photos(2)

Key Documents

ABC107

Sigma-Aldrich

Anti-KIF3A Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Kinesin-like protein KIF3A, Microtubule plus end-directed kinesin motor 3A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

species reactivity (predicted by homology)

baboon (based on 100% sequence homology), bovine (based on 100% sequence homology), chimpanzee (based on 100% sequence homology)

technique(s)

immunocytochemistry: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... KIF3A(11127)

General description

Kinesin-like protein KIF3 (KIF3A) is a member of the heteromeric family of kinesins and plays an important role in primary cilia formation. KIF3A functions as a microtubule-based anterograde translocator for membranous organelles and forms a heterotrimeric KIF3 complex with KIF3B or KIF3C and an associated protein, KAP3. Mutations in in KIF3A have been associated with cranial base growth retardation and dysmorphogenesis.

Specificity

Other homologies: Mouse, Rat, Chicken, Zebrafish, and Frog (92% sequence homology).

Immunogen

KLH-conjugated linear peptide corresponding to human KIF3A.

Application

Immunocytochemistry Analysis: A 1:500 dilution from a representative lot detected KIF3A in A431, HeLa, and NIH/3T3 cells.
Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional
This Anti-KIF3A Antibody is validated for use in ICC for the detection of KIF3A.

Quality

Evaluated by Western Blot in SH-SY5Y cell lysate.

Western Blot Analysis: 1 µg/mL of this antibody detected KIF3A in 10 µg of SH-SY5Y cell lysate.

Target description

~80 kDa observed

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
SH-SY5Y cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service