Skip to Content
Merck
All Photos(1)

Key Documents

AB9766

Sigma-Aldrich

Anti-Kv12.1 Antibody

Chemicon®, from rabbit

Synonym(s):

Elk1, KCNH8, ether-a-go-go like K + channel 3, Voltage gated K + channel subfamily H member 8

Sign Into View Organizational & Contract Pricing

Select a Size

200 μL
€583.00

€583.00


Estimated to ship on05 May 2025


Request a Bulk Order

Select a Size

Change View
200 μL
€583.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€583.00


Estimated to ship on05 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, rat

manufacturer/tradename

Chemicon®

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... KCNH8(131096)

Specificity

Recognizes Kv12.1 (Elk1, KCNH8, ether-a-go-go like K+ channel 3, Voltage gated K+ channel subfamily H member 8).
The immunogen sequence is 14 of 15 amino acids identical in mouse.

Immunogen

Purified peptide from amino acids 143-157 of rat Kv12.1 .

Application

Research Category
Neuroscience
Research Sub Category
Ion Channels & Transporters
This Anti-Kv12.1 Antibody is validated for use in WB for the detection of Kv12.1.
Western blot: 1:200 using ECL on rat cortex lysate.
Dilutions should be made using a carrier protein such as BSA (1-3%).

Optimal working dilutions must be determined by the end user.

Physical form

Affinity purified immunoglobulin. Lyophilized from phosphate buffered saline, pH 7.4, containing 1% BSA, and 0.025% sodium azide as a preservative. Reconstitute with 200 µL of sterile deionized water. Centrifuge antibody preparation before use (10,000 xg for 5 min).
ImmunoAffinity Purified

Storage and Stability

Maintain lyophilized material at -20°C for up to 12 months after date of receipt. After reconstitution maintain at -20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.

Analysis Note

Control
Included free of charge with the antibody is 40 µg of control antigen (lyophilized powder). The stock solution of the antigen can be made up using 100 µL of sterile deionized water. For negative control, preincubate 1 µg of peptide with 1 µg of antibody for one hour at room temperature. Optimal concentrations must be determined by the end user.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service