Skip to Content
Merck
All Photos(1)

Documents

AB9624

Sigma-Aldrich

Anti-Ephrin B phosphoTyr298 Antibody

Chemicon®, from rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat

manufacturer/tradename

Chemicon®

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

phosphorylation (pTyr298)

Gene Information

human ... EPHB1(2047)

Specificity

EphrinB, phosphoTyrosine 298. The antibody recognizes a protein of ~46 kDa corresponding to EphrinB, phosphoTyrosine 298 in lysates from rat testes. Immunolabeling is blocked by l-phosphatase treatment.

Immunogen

Synthetic peptide of amino acids surrounding the phosphoTyrosine 298 site of Xenopus EphrinB. Xenopus Tyrosine 298 is the homolog of mouse, rat and human Tyrosine 317 and chicken Tyrosine 305.

Application

Anti-Ephrin B phosphoTyr298 Antibody detects level of Ephrin B phosphoTyr298 & has been published & validated for use in WB.
Research Category
Neuroscience
Research Sub Category
Growth Cones & Axon Guidance
Western blot: 1:1,000. This antibody detects a strong band around 46kDa in rat testes lysate.

Optimal working dilutions must be determined by the end user.

Physical form

Affinity purified immunoglobulin. Liquid in 10 mM HEPES (pH 7.5), 150 mM NaCl, 100 μg/mL BSA and 50% glycerol.

Storage and Stability

Maintain at -20°C in undiluted for up to 6 months after date of receipt. Avoid repeated freeze/thaw cycles. Do not store in a self defrosting freezer.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service