Skip to Content
Merck
All Photos(2)

Key Documents

AB764P

Sigma-Aldrich

Anti-Procollagen Type III PIIINP Antibody

Chemicon®, from rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€683.00

€683.00


Estimated to ship on30 May 2025



Select a Size

Change View
100 μG
€683.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€683.00


Estimated to ship on30 May 2025


biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunohistochemistry: suitable
radioimmunoassay: suitable

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... PCOLCE(5118)

General description

Rabbit polyclonal antibodies against purified Procollagen Type III amino-terminal peptide (PIIINP). Antibody shows less than 0.1% reactivity with human and bovine Collagen Types I and III by radioimmunoassay.

Specificity

Recognizes Human type III procollagen: 100%. Human collagen type III, human collagen type I, human collagen type IV, human procollagen type I, human fibronectin and mouse laminin: <0.1%.

Application

Anti-Procollagen Type III PIIINP Antibody detects level of Procollagen Type III PIIINP & has been published & validated for use in ELISA, IH, RIA.
Immunohistochemistry: 1:40 dilution for immunofluorescent staining of frozen human and bovine skin and liver tissues.

Radioimmunoassay

ELISA: 100-1:200
Research Category
Cell Structure
Research Sub Category
ECM Proteins

Linkage

Replaces: AB764

Physical form

Format: Purified
ImmunoAffinity Purified
Lyophilized Solid

Storage and Stability

Maintain for 2 years at -20°C from date of shipment. Aliquot to avoid repeated freezing and thawing. For maximum recovery of product, centrifuge the original vial after thawing and prior to removing the cap.

Analysis Note

Control
Skin and liver tissues

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fraz Anjum et al.
Scientific reports, 7(1), 10291-10291 (2017-09-02)
Cell-based therapies have recently been the focus of much research to enhance skin wound healing. An important challenge will be to develop vehicles for cell delivery that promote survival and uniform distribution of cells across the wound bed. These systems

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service