Skip to Content
Merck
All Photos(1)

Key Documents

AB6039

Sigma-Aldrich

Anti-Vinculin Antibody

rabbit polyclonal

Synonym(s):

Metavinculin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

Product Name

Anti-Vinculin Antibody, from rabbit, purified by affinity chromatography

biological source

rabbit

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat, human, mouse

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... VCL(7414)

General description

Cell spreading and movement occur though the process of binding of cell surface integrin receptors to extracellular matrix adhesion molecules. In mammalian cells, vinculin is a membrane-cytoskeletal protein in focal adhesion plaques that is involved in linkage of integrin adhesion molecules to the actin cytoskeleton. More specifically, vinculin binds to talin at its N-terminus, which, in turn, binds to beta integrins. At its C-terminus, vinculin binds to actin, phospholipids, and paxillin-forming homodimers. The binding complex of vinculin to talin and actin serves to anchor actin filaments to the membrane, (Ezzell, 1997).

Vinculin′s ability to interact with integrins at the focal adhesion appears to be critical for control of cytoskeletal mechanics, cell spreading, and lamellipodia formation. Thus, vinculin appears to play a key role in shape control based on its ability to modulate focal adhesion structure and function.

Specificity

Predicted to cross react with monkey, pig, cow, and horse based on sequence homology.
The antibody recognizes an internal domain of human vinculin.

Immunogen

Epitope: Internal Domain
KLH-conjugated synthetic peptide corresponding to an internal domain of human Vinculin.

Application

Detect Vinculin using this Anti-Vinculin Antibody validated for use in WB.
Research Category
Cell Structure
Research Sub Category
Cytoskeletal Signaling

Cytoskeleton

Quality

Evaluated on a representative lot by Western blot on HepG2 cell lysate using Anti-Vinculin.

Target description

~ 124 kDa

Physical form

Antigen Affinity Purified
Purified in buffer containing 0.1 M Tris-Glycine, pH 7.4, 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
HepG2 cell lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cecilia Mancini et al.
Neurobiology of disease, 124, 14-28 (2018-11-06)
Spinocerebellar ataxia 28 is an autosomal dominant neurodegenerative disorder caused by missense mutations affecting the proteolytic domain of AFG3L2, a major component of the mitochondrial m-AAA protease. However, little is known of the underlying pathogenetic mechanisms or how to treat
Tolu Omokehinde et al.
Journal of bone and mineral research : the official journal of the American Society for Bone and Mineral Research, 37(2), 185-201 (2021-09-04)
Breast cancer cells frequently home to the bone marrow, where they encounter signals that promote survival and quiescence or stimulate their proliferation. The interleukin-6 (IL-6) cytokines signal through the co-receptor glycoprotein130 (gp130) and are abundantly secreted within the bone microenvironment.
Claudia Penna et al.
International journal of molecular sciences, 22(8) (2021-05-01)
Heart failure (HF) prevalence is increasing among the aging population, and the mortality rate remains unacceptably high despite improvements in therapy. Myocardial ischemia (MI) and, consequently, ischemia/reperfusion injury (IRI), are frequently the basis of HF development. Therefore, cardioprotective strategies to
Elisa Pozzi et al.
Scientific reports, 10(1), 20182-20182 (2020-11-21)
Short term treatment with low doses of glucocorticoid analogues has been shown to ameliorate neurological symptoms in Ataxia-Telangiectasia (A-T), a rare autosomal recessive multisystem disease that mainly affects the cerebellum, immune system, and lungs. Molecular mechanisms underlying this clinical observation
Rachelle W Johnson et al.
Nature cell biology, 18(10), 1078-1089 (2016-09-20)
Breast cancer cells frequently home to the bone marrow, where they may enter a dormant state before forming a bone metastasis. Several members of the interleukin-6 (IL-6) cytokine family are implicated in breast cancer bone colonization, but the role for

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service