Skip to Content
Merck
All Photos(1)

Key Documents

AB5787

Sigma-Aldrich

Anti-Calcium Channel β 2 Antibody, near CT

Chemicon®, from rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€358.00

€358.00


Estimated to ship on20 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
€358.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€358.00


Estimated to ship on20 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, mouse, rat

manufacturer/tradename

Chemicon®

technique(s)

western blot: suitable

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... CACNB2(783)

Specificity

beta2 calcium channel (Voltage-Dependant L-type Calcium Channel, beta2 subunit).

Immunogen

Epitope: near C-terminus
Synthetic peptide from C-terminus of rat beta2 calcium channel (SwissProt #Q8VGC3)

Application

Anti-Calcium Channel Antibody, β 2, near C-terminus is an antibody against Calcium Channel for use in WB.
Immunoblotting: 5-10 μg/mL. Reacts with the 68 kD protein.

Optimal working dilutions must be determined by the end user.
Research Category
Neuroscience
Research Sub Category
Ion Channels & Transporters

Signaling Neuroscience

Physical form

Format: Purified
Purified immunoglobulin. Liquid in PBS containing 0.08% sodium azide.

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 6 months after date of receipt. Avoid repeated freeze/thaw cycles.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service