Skip to Content
Merck
All Photos(1)

Key Documents

AB5779

Sigma-Aldrich

Anti-Striatin Antibody

serum, Chemicon®

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€385.00

€385.00


Estimated to ship on25 May 2025


Request a Bulk Order

Select a Size

Change View
100 μL
€385.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€385.00


Estimated to ship on25 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse, human, rat

manufacturer/tradename

Chemicon®

technique(s)

immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... STRN(6801)

General description

Striatin is found in all mammalian cells and may be involved in vesicular transport, dendrite growth and cellular signalling. It binds Protein phosphatase 2A (PP2A) A and C subunits and calmodulin and appears to modulate PP2A activity. It also binds the human homolog of the yeast protein, Mob1, which has been implicated in mitotic progression in yeast. It is a member of a family of proteins that also include SG2NA (striatin 3) and Zinedin (striatin 4).

Specificity

Reacts with Striatin. No cross reactivity with SG2NA.

Immunogen

Synthetic Striatin peptide.

Application

Anti-Striatin Antibody detects level of Striatin & has been published & validated for use in IP, WB & IC.
Research Category
Neuroscience
Research Sub Category
Synapse & Synaptic Biology
Western blot: 1:500-1:1,000. The antibody reacts with the ~110 kDa striatin protein in 293 cell lysate.

Immunocytochemistry: 1:500

Immunoprecitation: 3 μL recommended as starting point.

Optimal working dilutions must be determined by the end user.

Physical form

Rabbit serum. Liquid containing 40% glycerol and 0.02% sodium azide.

Storage and Stability

Maintain at -20°C in undiluted for up to 12 months after date of receipt. For long-term storage, may be stored at -80°C. Avoid repeated freeze/thaw cycles at temperatures below -20°C.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sean E Thatcher et al.
Biology of sex differences, 6, 12-12 (2015-07-02)
Abdominal aortic aneurysms (AAAs) occur predominately in males. However, AAAs in females have rapid growth rates and rupture at smaller sizes. Mechanisms contributing to AAA progression in females are undefined. We defined effects of ovariectomy, with and without 17-β estradiol

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service