Skip to Content
Merck
All Photos(1)

Key Documents

AB5669

Sigma-Aldrich

Anti-Spinophilin Antibody

Chemicon®, from rabbit

Synonym(s):

Neurabin-II

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€614.00

€614.00


Estimated to ship on25 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
€614.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€614.00


Estimated to ship on25 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat

manufacturer/tradename

Chemicon®

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... PPP1R9B(84687)

Specificity

Spinophilin

Immunogen

Synthetic peptide from rat Spinophilin.

Application

Anti-Spinophilin Antibody detects level of Spinophilin & has been published & validated for use in WB.
Research Category
Neuroscience
Research Sub Category
Synapse & Synaptic Biology

Neuronal & Glial Markers
Western blot: 0.05 - 0.1 μg/mL using ECL on rat brain lysate. Reacts with a band of ~140 kDa.

Optimal working dilutions must be determined by the end user.

Physical form

Affinity purified immunoglobulin. Liquid in 0.02M phosphate buffer containing 0.25M NaCl, pH 7.6 with 0.1% sodium azide.

Storage and Stability

Maintain at 2-8°C in undiluted for up to 6 months after date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Micah A Shelton et al.
Biological psychiatry, 78(6), 374-385 (2015-03-31)
Microtubule-associated protein 2 (MAP2) is a neuronal protein that plays a role in maintaining dendritic structure through its interaction with microtubules. In schizophrenia (Sz), numerous studies have revealed that the typically robust immunoreactivity (IR) of MAP2 is significantly reduced across
Zhiping Mi et al.
Neurobiology of aging, 55, 159-166 (2017-03-06)
Precuneus (PreC) cortex is affected with amyloid plaques early in Alzheimer's disease (AD), and this pathology may be associated with alterations in PreC synapses and cognitive impairment. We quantified the spinophilin-immunoreactive (ir) dendritic spine density and the intensity of spinophilin
Steven R Boikess et al.
The European journal of neuroscience, 28(10), 2099-2107 (2008-12-03)
Structural studies have shown that chronic regimens of psychostimulants increase dendritic spine number in the rat striatum. The present study used Western blotting and radioimmunocytochemistry to examine psychostimulant-induced changes in the levels of spinophilin, a protein found abundantly in dendritic
Kisho Obi-Nagata et al.
Science advances, 9(23), eade5973-eade5973 (2023-06-09)
Human genetics strongly support the involvement of synaptopathy in psychiatric disorders. However, trans-scale causality linking synapse pathology to behavioral changes is lacking. To address this question, we examined the effects of synaptic inputs on dendrites, cells, and behaviors of mice
Melanie J Grubisha et al.
Proceedings of the National Academy of Sciences of the United States of America, 118(49) (2021-12-02)
Normally, dendritic size is established prior to adolescence and then remains relatively constant into adulthood due to a homeostatic balance between growth and retraction pathways. However, schizophrenia is characterized by accelerated reductions of cerebral cortex gray matter volume and onset

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service