Skip to Content
Merck
All Photos(1)

Key Documents

AB5666P

Sigma-Aldrich

Anti-Neurite Outgrowth Inhibitor A/B/C Antibody

Chemicon®, from rabbit

Synonym(s):

Nogo-A/B/C

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, mouse, rat

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... RTN4(57142)

Specificity

Recognizes human Neurite Outgrowth Inhibitor Protein A/B/C (Nogo-A/B/C). The immunogen peptide is within the 25 amino acid region identified for the inhibition of neurite outgrowth (Chen et al. 2000; GrandPre et al. 2000; Goldberg & Barres 2000; Prinijha et al. 2000; Tessier-Lavigne & Goodman 2000; Nagase et al. 1998).The immunogen shows no significant sequence homology with other known proteins.

SPECIES REACTIVITIES: The immunogen peptide is 92% conserved in rat Nogo or rat foocen-m1.2 protein (Chen et al. 2000; GrandPre et al. 2000; Goldberg & Barres 2000; Prinijha et al. 2000; Tessier-Lavigne & Goodman 2000; Nagase et al. 1998). Reactivity with other species has not been confirmed.

Immunogen

A 13 amino acid peptide between the putative 2nd-3rd transmembrane domain of human Nogo-A (Chen et al. 2000; GrandPre et al. 2000; Goldberg & Barres 2000; Prinijha et al. 2000; Tessier-Lavigne & Goodman 2000; Nagase et al. 1998).

Application

Anti-Neurite Outgrowth Inhibitor A/B/C Antibody is an antibody against Neurite Outgrowth Inhibitor A/B/C for use in ELISA & WB.
Research Category
Neuroscience
Research Sub Category
Neurochemistry & Neurotrophins

Growth Cones & Axon Guidance
Western blot: 1-10 μg/mL using ECL. The calculated molecular weight of Nogo-A is approximately 135 kDa. However a molecular weight of approximately 180 kDa has been reported for full length Nogo-A (Chen et al. 2000; GrandPre et al. 2000; Goldberg & Barres 2000; Prinijha et al. 2000; Tessier-Lavigne & Goodman 2000; Nagase et al. 1998).



ELISA: 1:10,000-1:100,000 using 50-100 ng control peptide per well.

Optimal working dilutions must be determined by the end user.

Physical form

Liquid in PBS containing 0.1% BSA and 0.05% sodium azide.

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service