Skip to Content
Merck
All Photos(1)

Key Documents

AB5644P

Sigma-Aldrich

Anti-Fatty Acid Amide Hydrolase Antibody

Chemicon®, from rabbit

Synonym(s):

FAAH

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
€421.00

€421.00


Estimated to ship on15 May 2025


Request a Bulk Order

Select a Size

Change View
50 μG
€421.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€421.00


Estimated to ship on15 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, rat

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... FAAH(2166)

Specificity

Recognizes human Fatty Acid Amide Hydrolase (FAAH). The immunogen shows no significant sequence homology with other hydrolases

SPECIES REACTIVITIES: The immunogen peptide is 100% conserved in rat, 94% in mouse and porcine and 58% in chicken. Reactivity with other species has not been confirmed.

Immunogen

A 17 amino acid peptide sequence near the N-terminus of human FAAH (Patricelli et al. 1998; Maccarone et al. 1998; Cravatt et al. 1996; Giang et al. 1997; Thomas et al. 1997; Wan et al. 1998).

Application

This Anti-Fatty Acid Amide Hydrolase Antibody is validated for use in ELISA, WB for the detection of Fatty Acid Amide Hydrolase.
Western blot: 1-10 μg/mL using ECL.

ELISA: 1:10,000-1:100,000 using 50-100 ng control peptide (Catalog Number AG376) per well.

Optimal working dilutions must be determined by the end user.

Physical form

Affinity purified immunoglobulin. Liquid in 100 mM Tris, pH 7.5, containing 0.2% BSA and 0.05% sodium azide.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service