Skip to Content
Merck
All Photos(1)

Key Documents

AB5403

Sigma-Aldrich

Anti-NMDAR2B Antibody, phosphoTyr 1472

Chemicon®, from rabbit

Synonym(s):

Anti-DEE27, Anti-EIEE27, Anti-GluN2B, Anti-MRD6, Anti-NMDAR2B, Anti-NR2B, Anti-NR3, Anti-hNR3

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€737.00

€737.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
100 μL
€737.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€737.00


Estimated to ship on28 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat

manufacturer/tradename

Chemicon®

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

Specificity

NMDAR2B, phosphoTyr1472. The antibody recognizes a protein of ~180 kDa in Western blot. The labeling of the NMDAR2B protein in Western blots of rat brain is blocked by the phosphopeptide but not by the dephosphopeptide. Two additional lower molecular weight bands are present in Western blots of rat brain.

Immunogen

Epitope: phosphoTyr 1472
Synthetic peptide corresponding to amino acids surrounding the phosphoTyr1472 of rat NMDAR2B.

Application

Anti-NMDAR2B Antibody, phosphoTyr 1472 is an antibody against NMDAR2B for use in WB.
Research Category
Neuroscience
Research Sub Category
Neurotransmitters & Receptors
Western blot: 1:1,000

Optimal working dilutions must be determined by the end user.

Physical form

Affinity purified immunoglobulin. Liquid in 10 mM HEPES (pH 7.5), 150 mM NaCl with 100 μg/mL BSA, 50% glycerol and 0.09% sodium azide.

Storage and Stability

Maintain at -20°C in undiluted for up to 6 months after date of receipt. Avoid repeated freeze/thaw cycles. Do not store in a self defrosting freezer.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tara Canonica et al.
Frontiers in behavioral neuroscience, 18, 1428146-1428146 (2024-07-26)
Trisomy of human chromosome 21 (Hsa21) results in a constellation of features known as Down syndrome (DS), the most common genetic form of intellectual disability. Hsa21 is orthologous to three regions in the mouse genome on mouse chromosome 16 (Mmu16)
Youn Hee Jee et al.
Genetics in medicine : official journal of the American College of Medical Genetics, 22(8), 1329-1337 (2020-04-29)
Impaired function of gonadotropin-releasing hormone (GnRH) neurons can cause a phenotypic spectrum ranging from delayed puberty to isolated hypogonadotropic hypogonadism (IHH). We sought to identify a new genetic etiology for these conditions. Exome sequencing was performed in an extended family
Sofia Lopes et al.
Proceedings of the National Academy of Sciences of the United States of America, 113(26), E3755-E3763 (2016-06-09)
Exposure to chronic stress is frequently accompanied by cognitive and affective disorders in association with neurostructural adaptations. Chronic stress was previously shown to trigger Alzheimer's-like neuropathology, which is characterized by Tau hyperphosphorylation and missorting into dendritic spines followed by memory
Sean McKay et al.
Cell reports, 25(4), 841-851 (2018-10-26)
The GluN2 subtype (2A versus 2B) determines biophysical properties and signaling of forebrain NMDA receptors (NMDARs). During development, GluN2A becomes incorporated into previously GluN2B-dominated NMDARs. This "switch" is proposed to be driven by distinct features of GluN2 cytoplasmic C-terminal domains
Jun-Wu Wang et al.
Anesthesia and analgesia, 135(4), 865-876 (2022-07-13)
The number of patients with diabetic neuropathic pain (DNP) continues to increase, but available treatments are limited. This study aimed to examine the influence of reactive oxygen species (ROS)-thioredoxin-interacting protein (TXNIP)-NOD-like receptor protein 3 (NLRP3)- N -methyl-D-aspartic acid receptor 2B

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service