Skip to Content
Merck
All Photos(2)

Key Documents

AB5336P

Sigma-Aldrich

Anti-α-Synuclein (SNCA) Antibody

CHEMICON®, sheep polyclonal

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
€536.00

€536.00


Estimated to ship on23 May 2025

A recombinant, preservative-free antibody is available for your target. Try ZRB1134

Request a Bulk Order

Select a Size

Change View
50 μG
€536.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€536.00


Estimated to ship on23 May 2025

A recombinant, preservative-free antibody is available for your target. Try ZRB1134

Request a Bulk Order

Product Name

Anti-Synuclein α Antibody, Chemicon®, from sheep

biological source

sheep

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat, mouse, human

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable (paraffin)

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... SNCA(6622)

Immunogen

Synthetic peptide corresponding amino acids 108-120 of human alpha synuclein.

Application

Anti-Synuclein Antibody, α is an antibody against Synuclein for use in IH(P).
Immunohistochemistry: 1:1,000 on frozen or paraffin sections.

Optimal working dilutions must be determined by the end user.

Linkage

Replaces: 04-1053

Physical form

Affinity purified immunoglobulin. Lyophilized. Reconstitute with 50 μL of sterile distilled water. Centrifuge to remove any residue. Glycerol (1:1) can be added for additional stability.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mohamed-Bilal Fares et al.
Proceedings of the National Academy of Sciences of the United States of America, 113(7), E912-E921 (2016-02-04)
Lewy bodies (LBs) are intraneuronal inclusions consisting primarily of fibrillized human α-synuclein (hα-Syn) protein, which represent the major pathological hallmark of Parkinson's disease (PD). Although doubling hα-Syn expression provokes LB pathology in humans, hα-Syn overexpression does not trigger the formation
Avik Roy et al.
Scientific reports, 6, 22067-22067 (2016-02-27)
Ankyrin-rich BTB/POZ domain containing protein-2 or BPOZ-2, a scaffold protein, has been recently shown to control the degradation of many biological proteins ranging from embryonic development to tumor progression. However, its role in the process of neuronal diseases has not
Ayse Ulusoy et al.
The European journal of neuroscience, 32(3), 409-422 (2010-08-14)
Lewy bodies, which are a pathological hallmark of Parkinson's disease, contain insoluble polymers of alpha-synuclein (alphasyn). Among the different modifications that can promote the formation of toxic alphasyn species, C-terminal truncation is among the most abundant alterations in patients with
Martial Kamdem Mbefo et al.
The Journal of biological chemistry, 290(15), 9412-9427 (2015-02-07)
Although α-synuclein (α-syn) phosphorylation has been considered as a hallmark of sporadic and familial Parkinson disease (PD), little is known about the effect of PD-linked mutations on α-syn phosphorylation. In this study, we investigated the effects of the A30P, E46K
Cristiane Latge et al.
The Journal of biological chemistry, 290(33), 20527-20540 (2015-07-08)
Cerebral dopamine neurotrophic factor (CDNF) is a promising therapeutic agent for Parkinson disease. As such, there has been great interest in studying its mode of action, which remains unknown. The three-dimensional crystal structure of the N terminus (residues 9-107) of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service