Skip to Content
Merck
All Photos(4)

Key Documents

AB5152

Sigma-Aldrich

Anti-Calcium Channel Antibody, Voltage Gated α 1A

Chemicon®, from rabbit

Synonym(s):

P/Q-type

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat, mouse

manufacturer/tradename

Chemicon®

technique(s)

immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CACNA1A(773)

Specificity

Recognizes Cav2.1 (alpha 1A subunit, P/Q-type of voltage-gated calcium channel, Cacna1a). Does not cross react with any other calcium channel proteins tested so far.

Immunogen

Purified peptide CNA1 from 865-881 of alpha 1A subunit of rat brain voltage-gated calcium channel (Starr et al. 1991) (Accession P54282) containing N-terminal lysine and tyrosine, conjugated to KLH with gluteraldehyde (Westenbroek et al. 1995).

Application

Anti-Calcium Channel Antibody, Voltage Gated α 1A is an antibody against Calcium Channel for use in IC, IP & WB.
In preparing full-length protein samples for Western blot or immunoprecipitation, the procedures should be performed at 4°C, with the following protease inhibitor mixture: pepstatin A (1 μg/mL), leupeptin (1 μg/mL), aprotinin (1 μg/mL), Pefabloc SC (0.2 mM), benzamidine (0.1 mg/mL), and calpain inhibitors I and II (8 μg/mL each)

Immunoprecipitation: 3 μg/mL

Immunocytochemistry: 5 μg/mL (1:60). Recommend subsequent staining with a labeled

second antibody.

Western blot: 1.5 μg/mL (1:200) using ECL

Dilutions should be made using a carrier protein such as BSA (1-3%)



Optimal working dilutions must be determined by the end user.

Storage and Stability

Maintain lyophilized material at -20°C for up to 12 months after date of receipt. After reconstitution add glycerol (ASC gade or better at a ratio of 1:1 and maintain at -20°C for up to 6 months. Avoid repeated freeze/thaw cycles.

Analysis Note

Control
CONTROL ANTIGEN: Included free of charge with the antibody is 20 μg of control antigen (lyophilized powder in phosphate buffered saline, pH 7.4, containing 5% sucrose and 0.025% sodium azide). The stock solution of the antigen can be made up using 100 μL of sterile deionized water. For negative control, preincubate 1 μg of peptide with 10 μg of antibody for one hour at room temperature. Optimal concentrations must be determined by the end user.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Arn M J M van den Maagdenberg et al.
Neuron, 41(5), 701-710 (2004-03-09)
Migraine is a common, disabling, multifactorial, episodic neurovascular disorder of unknown etiology. Familial hemiplegic migraine type 1 (FHM-1) is a Mendelian subtype of migraine with aura that is caused by missense mutations in the CACNA1A gene that encodes the alpha(1)
Christoph Limbach et al.
Proceedings of the National Academy of Sciences of the United States of America, 108(31), E392-E401 (2011-06-30)
The protein machinery of neurotransmitter exocytosis requires efficient orchestration in space and time, for speed and precision of neurotransmission and also for synaptic ontogeny and plasticity. However, its spatial organization in situ is virtually unknown. Aczonin/Piccolo is a putative organizer

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service