Skip to Content
Merck
All Photos(1)

Key Documents

AB5018

Sigma-Aldrich

Anti-Glutamate (No Glutaraldehyde) Antibody

Chemicon®, from rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

2000 ASSAYS
€600.00

€600.00


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
2000 ASSAYS
€600.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€600.00


Estimated to ship on29 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity (predicted by homology)

all

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable

shipped in

wet ice

target post-translational modification

unmodified

Specificity

Glutamate (L-Glutamate). The antibody has been calibrated against a spectrum of antigens to assure hapten selectivity and proper affinity. No measurable cross-reactivity (<1:1000) against glutamate in peptides or proteins. No measurable glutaraldehyde-fixed tissue cross-reactivity (<1:1000) against L-alanine, gamma-aminobutyrate, 1-amino-4-guanidobutane (AGB), D/L-arganine, D/L-aspartate, L-citrulline, L-cysteine, D/L-glutamine, glutathione, glycine, L-lysine, L-ornithine, L-serine, taurine, L-threonine, L-tryptophan, L-tyrosine. Modest cross-reactivity (1:20) against D-glutamate. Significant cross-reactivity (1:8) against free NAAG in competition assays (NAAG is not retained in glutaraldehyde-fixed tissue).

Immunogen

Glutamate-glutaraldehyde

Application

Immunohistochemistry using silver-intensified immunogold or fluorescence (see recommended protocol). Samples should be fixed with 0.1% - 2.5% glutaraldehyde for optimum detection.

*This antibody has also been used and found to work with a zero-low glutaraldehyde / high paraformaldehyde fixation (4% paraformaldehyde in 0.1M phosphate buffer / 3% sucrose fixative ). The minimum glutaraldehyde concentration for AB5018 is 0.00%. See protocol that follows. Performance is good with frozen sections, Vibratome sections and tissue culture formats, when penetrating reagents such as 0.3% Triton X-100 are used.

Optimal working dilutions must be determined by the end user.

DILUTION: Prepare enough of the AB5018 for your days use by diluting 100X with 1% GSPBT.
Research Category
Neuroscience
Research Sub Category
Neurotransmitters & Receptors

Neuroinflammation & Pain
This Anti-Glutamate (No Glutaraldehyde) Antibody is validated for use in IH for the detection of Glutamate (No Glutaraldehyde).

Physical form

Format: Purified
IgG fraction in sterile 0.1M phosphate buffer. No preservative.

Storage and Stability

Maintain stock at 2-8°C in undiluted aliquots for up to 6 months. This stock is extremely stable under normal use and routine storage at 2-8°C. Do not freeze.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Md Abdul Alim et al.
Cellular & molecular immunology (2020-04-22)
Mast cells are emerging as players in the communication between peripheral nerve endings and cells of the immune system. However, it is not clear the mechanism by which mast cells communicate with peripheral nerves. We previously found that mast cells
E V Megalou et al.
The Biological bulletin, 216(2), 103-112 (2009-04-16)
The escape swim response of the marine mollusc Tritonia diomedea is a well-established model system for studies of the neural basis of behavior. Although the swim neural network is reasonably well understood, little is known about the transmitters used by
N Schizas et al.
Scandinavian journal of medicine & science in sports, 20(2), 208-215 (2009-05-09)
Elevated levels of the neurotransmitter glutamate and the presence of its receptor, N-methyl-d-aspartate receptor type 1 (NMDAR1), have been established in patients with tendinopathy, i.e. chronic tendon pain and degeneration. However, whether NMDAR1 is up- or down-regulated in tendinopathy and
Puneet Bagga et al.
Scientific reports, 8(1), 2883-2883 (2018-02-15)
Glutamate Chemical Exchange Saturation Transfer (GluCEST) MRI is a recently developed technique to image glutamate. In the present study, we evaluated the reproducibility and background contamination to the GluCEST and source of the GluCEST changes in a mouse model of
Réka Horváth et al.
Brain structure & function, 225(8), 2563-2575 (2020-09-21)
Perception and processing of chemical cues are crucial for aquatic gastropods, for proper elaboration of adaptive behavior. The pond snail, Lymnaea stagnalis, is a model species of invertebrate neurobiology, in which peripheral sensory neurons with different morphology and transmitter content

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service