Skip to Content
Merck
All Photos(1)

Key Documents

AB3841

Sigma-Aldrich

Anti-Connexin 43 Antibody, phospho-specific (Ser368)

Chemicon®, from rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€390.00

€390.00


Estimated to ship on18 May 2025


Request a Bulk Order

Select a Size

Change View
100 μL
€390.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€390.00


Estimated to ship on18 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

hamster, mouse, rat, monkey, human

manufacturer/tradename

Chemicon®

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

phosphorylation (pSer368)

Gene Information

human ... GJA1(2697)

Specificity

Recognizes Ser368 phosphorylated Connexin 43. Connexin 43 (Cx43) is a member of the large family of gap junction proteins. Connexins assemble as a hexamer and are transported to the plasma membrane to create a hemichannel that can associate with hemichannels on nearby cells to create cell-to-cell channels. Clusters of these channels assemble to make gap junctions. Gap-junction communication is important in development and regulation of cell growth.

Immunogen

Epitope: phospho-specific (Ser368)
KLH-conjugated, synthetic phospho-peptide corresponding to residues surrounding Ser368 of human Connexin 43.

Application

Research Category
Cell Structure
Research Sub Category
Adhesion (CAMs)
This Anti-Connexin 43 Antibody, phospho-specific (Ser368) is validated for use in WB for the detection of Connexin 43.
Western blotting 1:1,000 (for best results, incubate membrane with diluted antibody in 5% BSA, 1X TBS, 0.1% Tween-20 at 4°C with gentle shaking, overnight)

Optimal working dilutions must be determined by the end user.

Physical form

Protein A and peptide affinity purified immunoglobulin. Liquid in 10 mM sodium HEPES, pH 7.5, 150 mM NaCl, 100 μg/mL BSA, with 50% glycerol.

Storage and Stability

Maintain at -20ºC for 12 months after date of receipt. Do not aliquot.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

David Johansen et al.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology, 28(1), 103-114 (2011-08-26)
Gap junction intercellular communication (GJIC) and hemichannel permeability may have important roles during an ischemic insult. Our aim was to evaluate the effect of ischemia on gap junction channels and hemichannels. We used neonatal rat heart myofibroblasts and simulated ischemia
Vladimir Akoyev et al.
Cellular signalling, 19(5), 958-967 (2007-01-11)
We have previously reported that protein kinase C gamma (PKC-gamma) is activated by phorbol-12-myristate-13-acetate (TPA) and that this causes PKC-gamma translocation to membranes and phosphorylation of the gap junction protein, connexin 43 (Cx43). This phosphorylation, on S368 of Cx43, causes
Ronald Ng et al.
JCI insight, 5 (2019-06-14)
Arrhythmogenic cardiomyopathy (ACM) is an inherited disorder with variable genetic etiologies. Here we focused on understanding the precise molecular pathology of a single clinical variant in DSP, the gene encoding desmoplakin. We initially identified a novel missense desmoplakin variant (p.R451G)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service