Skip to Content
Merck
All Photos(1)

Key Documents

AB3749

Sigma-Aldrich

Anti-mGCM1 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

GCM motif protein 1, Glial cells missing homolog 1, chorion-specific transcription factor GCMa, glial cells missing homolog a

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

western blot: suitable

input

sample type pancreatic stem cell(s)
sample type neural stem cell(s)

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... GCM1(8521)

General description

GCM1 is a DNA binding protein with a GCM motif (glial cell missing motif) that is a key transcription factor for determining the fate of stem cells within the nervous system, in addition to being critical for placental development. This protein binds to the GCM motif (A/G)CCCGCAT, a novel sequence among known targets of DNA binding proteins. The N-terminal DNA-binding domain confers the unique DNA binding activity of this protein. Current studies indicate that the introduction of the mouse GCM1 gene in cultured cells from embryonic brains resulted in the induction of an astrocyte lineage.

Specificity

Comparison of sequence homology suggests that this antibody will also react in mouse and rat; however, reactivity to other species has not been tested.
This antibody is generated from rabbits immunized with a KLH conjugated synthetic peptided selected from the C-terminal region mouse GCM1.

Immunogen

Epitope: C-terminus
Synthetic GCM1 peptide, a.a. 401-418.

Application

Research Category
Stem Cell Research
Research Sub Category
Neural Stem Cells
This Anti-mGCM1 Antibody is validated for use in WB for the detection of mGCM1.

Quality

Routinely evaluated by western blot on HEK-293 lysates.

Western Blotting: Recommended working dilutions are 1:1000 – 1:2500.

Target description

50 kDa

Physical form

Peptide affinity purified
Purified rabbit polyclonal antibody in buffer containing PBS with 0.09% sodium azide.

Storage and Stability

Maintain refrigerated at 2-8°C in undiluted aliquots for up to 12 months. Avoid repeat freeze/thaw cycles.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service