Skip to Content
Merck
All Photos(1)

Key Documents

AB3613

Sigma-Aldrich

Anti-Caspase 12 Antibody, NT

Chemicon®, from rabbit

Synonym(s):

Anti-CASP-12, Anti-CASP12P1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€472.00

€472.00


Estimated to ship on16 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
€472.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€472.00


Estimated to ship on16 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, mouse, rat

manufacturer/tradename

Chemicon®

technique(s)

western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

General description

Three distinct signaling pathways lead to apoptosis. The death receptor and mitochondrion pathways are the two mains, which involve respectively caspase-8 and caspase-9. The endoplasmic reticulum stress is the third apoptotic pathway and involves caspase-12. Localized to the ER but not to cytoplasm or mitochondria, caspase-12 is activated by ER stress, including disruption of ER calcium homeostasis. Caspase-12 is co-localized to the ER with several proteins that are involved in Alzheimer′s disease including g-secretase presenilin and b-amyloid precursor protein (APP). Caspase-12 mediates ER stress-induced apoptosis as well as cytotoxicity induced by amyloid-b. Caspase-12 is ubiquitously expressed in mouse tissues.

Specificity

Antibody recognizes caspase 12 pro-protein in mouse extracts of normal and apoptotic cells. Caspase 12 is predicted to have a large >120aa long prodomain region. Thus this antibody recognizes only the proforms of the enzyme and not the active subunits. Uncleaved (proform) caspase 12 is prominently detected in many tissues. Active forms are difficult to detect expect in transfect cell systems. {Kalai, M et al (2003) 162:457-467}.

Immunogen

Epitope: N-terminus
Peptide corresponding to aa 2-17 of mouse caspase-12.

Application

Research Category
Apoptosis & Cancer

Metabolism
Research Sub Category
Caspases

Enzymes & Biochemistry
This Anti-Caspase 12 Antibody, N-terminus is validated for use in WB for the detection of Caspase 12.
Western blot: 1:500 to 1:1000. Detects a band at ~53-49 kDa. in normal tissue extracts. Caspase 12 is a stress induced caspase localized to the ER and activated upon ER stress. Active process forms are difficult to detect in most non-transfected systems. Not all cells produce caspase 12; mouse L929 fibrosacrcoma cells are an excellent positive control line.

Mouse brain tissue lysate can be used as a positive control as well. Caspase 12 is associated with ER membranes and not in cytoplasm or mitochondria. Caspase 12 mediates cytotoxixity induced by amyloid beta.

Immunohistochemistry: 10μg/mL

Optimal working dilutions must be determined by end user.

Physical form

Affinity purified immunoglobulin. Liquid in PBS containing 0.02% sodium azide.
Format: Purified

Storage and Stability

Maintain at 4°C for up to 6 months.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Placido Rojas-Franco et al.
Journal of Zhejiang University. Science. B, 22(7), 603-608 (2021-07-17)
Arthrospira maxima (Spirulina) is a cyanobacterium which is considered a nutraceutical because it has antioxidant, anti-inflammatory, and cytoprotective properties in different renal disease models (Rodriguez-Sánchez et al., 2012; Aziz et al., 2018; Memije-Lazaro et al., 2018). The therapeutic effects are
Alejandra Paola Torres-Manzo et al.
Oxidative medicine and cellular longevity, 2018, 2089404-2089404 (2018-05-11)
Thyroid hormones (TH) are essential for hippocampal neuronal viability in adulthood, and their deficiency causes hypothyroidism, which is related to oxidative stress events and neuronal damage. Also, it has been hypothesized that hypothyroidism causes a glucose deprivation in the neuron.
Plácido Rojas-Franco et al.
Renal failure, 41(1), 1001-1010 (2019-11-19)
Acute exposure to mercury chloride (HgCl2) causes acute kidney injury (AKI). Some metals interfere with protein folding, leading to endoplasmic reticulum stress (ERS), and the activation of cell death mechanisms, but in the case of mercury, there is no knowledge

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service