Skip to Content
Merck
All Photos(1)

Key Documents

AB3609

Sigma-Aldrich

Anti-Smac/Diablo Antibody

Chemicon®, from rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€385.00

€385.00


Estimated to ship on22 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
€385.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€385.00


Estimated to ship on22 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, mouse, rat

manufacturer/tradename

Chemicon®

technique(s)

western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... DIABLO(56616)

General description

The inhibitor of apoptosis (IAP) proteins regulate programmed cell death by inhibiting members of the caspase family of enzymes. A novel mammalian protein that binds to IAPs and neutralizes their inhibitory effect on caspases has been designated Smac/DIABLO. This is a mitochondrial protein that is released along with cytochrome c during apoptosis and activates the cytochrome c/Apaf-1/caspase-9 pathway. Analysis of the structural basis of Smac/DIABLO reveals that the N-terminal amino acids are required for binding of Smac/DIABLO to IAPs and activation of caspases. Smac/DIABLO is expressed in a variety of human and mouse tissues.

Specificity

Detects Smac/DIABLO in an approximately 25 kDa band by western blot.

Immunogen

Peptide corresponding to aa 225-239 (EERAESEQEAYLRED) of human Smac/DIABLO.

Application

This Anti-Smac/Diablo Antibody is validated for use in WB for the detection of Smac/Diablo.
Western blot: 1:500 to 1:1000

Human heart tissue lysate can be used as a positive control.

Optimal working dilutions must be determined by end user.

Physical form

Format: Purified

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

APOLLON protein promotes early mitotic CYCLIN A degradation independent of the spindle assembly checkpoint.
Kikuchi, R; Ohata, H; Ohoka, N; Kawabata, A; Naito, M
The Journal of Biological Chemistry null
Zhaoqing Yang et al.
Arteriosclerosis, thrombosis, and vascular biology, 29(8), 1213-1219 (2009-05-16)
Increasing evidence suggests that chronic inflammation contributes to atherogenesis, and that acute inflammatory events cause plaque rupture, thrombosis, and myocardial infarction. The present studies examined how inflammatory factors, such as interferon-gamma (IFNgamma), cause increased sensitivity to apoptosis in vascular lesion
Zhaoqing Yang et al.
Journal of vascular research, 44(6), 483-494 (2007-07-28)
The inappropriate survival of cells in the neointima contributes to atherosclerotic plaque progression, while apoptosis in the fibrous cap of lesions contributes to myocardial infarction and stroke. Prior genomic-scale transcript profiling of human carotid artery plaque cells with known sensitivity

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service