Skip to Content
Merck
All Photos(1)

Key Documents

AB3579

Sigma-Aldrich

Anti-BimEL (pS69) human / (pS65) rat Antibody

Chemicon®, from rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€587.00

€587.00


Estimated to ship on18 May 2025


Request a Bulk Order

Select a Size

Change View
100 μL
€587.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€587.00


Estimated to ship on18 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat, human

species reactivity (predicted by homology)

mouse (100% homologous)

manufacturer/tradename

Chemicon®

technique(s)

western blot: suitable

UniProt accession no.

shipped in

dry ice

target post-translational modification

phosphorylation (pSer69)

Gene Information

human ... BCL2L11(10018)

General description

Bim (bcl-2-interacting mediator of cell death) is a proapoptotic member of the Bcl-2 family that shares only the BH3 domain with this family. There are three isoforms of Bim: BimEL, BimL, and BimS. Bim is involved in regulating the intrinsic mitochondrial apoptotic pathway by inducing cytochrome c release, which in turn, activates caspase-9 and then caspase-3. Bim also plays a critical role in central and peripheral deletion of T lymphocytes and in controlling B cell homeostasis and activation. BimEL, the long isoform of Bim, is a ~28 kDa protein that is predominantly expressed in T and B cells and is activated by ERK1/2 pathway. The activation of BimEL by ERK1/2 promotes its phosphorylation on serine 65 (serine 69 in human), targeting it for degradation via the proteasome.

Specificity

Rat BimEL when phosphorylated at serine 65. Human BimEL when phosphorylated at serine 69. Mouse BimEL (100% conserved) has not been tested, but is expected to react.

Immunogen

The antiserum was produced against a chemically synthesized phosphopeptide derived from the region of rat BimEL that contains serine 65 (serine 69 in the human sequence). The sequence is conserved in human and mouse.

Application

Anti-BimEL (pS69) human / (pS65) rat Antibody is an antibody against BimEL (pS69) human / (pS65) rat for use in WB.
For Western blotting applications, we recommend using the antibody at a 1:1000 dilution.
Research Category
Apoptosis & Cancer
Research Sub Category
BCL2 & Inhibition

Quality

Purified from rabbit serum by sequential epitope-specific chromatography. The antibody has been negatively preadsorbed using a non-phosphopeptide corresponding to the site of phosphorylation to remove antibody that is reactive with non-phosphorylated Bim. The final product is generated by affinity chromatography using a BimEL-derived peptide that is phosphorylated at serine 65 (serine 69 in the human sequence).

Physical form

Dulbecco′s phosphate buffered saline (without Mg2+ and Ca2+), pH 7.3 (+/- 0.1), 50% glycerol, with 1.0 mg/mL BSA (IgG, protease free) as a carrier. 0.05% sodium azide

Storage and Stability

Store at -20°C. We recommend a brief centrifugation before opening to settle vial contents. Then, apportion into working aliquots and store at -20°C. For shipment or short-term storage (up to one week), 2-8°C is sufficient.

Analysis Note

Control
Hek293T co-expressing rat BimEL and active MEKK1 (MEKK1δ).

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

ID4 regulates mammary gland development by suppressing p38MAPK activity.
Dong, J; Huang, S; Caikovski, M; Ji, S; McGrath, A; Custorio, MG; Creighton et al.
Development null
Raquel Delgado et al.
Cancers, 13(12) (2021-07-03)
Chronic lymphocytic leukemia (CLL) is a common form of leukemia with a heterogeneous clinical course that remains incurable due to the development of therapy resistance. In lymph node proliferation centers, signals from the microenvironment such as CD40 ligation through interaction

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service