Skip to Content
Merck

AB3573

Anti-BCL-XL (pS62) Antibody

Chemicon®, from rabbit

Synonym(s):

Bcl-2-binding component 6, Bcl-XL/Bcl-2-associated death promoter, Bcl-2-like 8 protein, BAD, BBC6, BCL2L8

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Clone:
polyclonal
Species reactivity:
human
Application:
WB
Citations:
8
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

species reactivity (predicted by homology)

rat (100% homologous), mouse (100% homologous)

manufacturer/tradename

Chemicon®

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

phosphorylation (pSer62)

Gene Information

human ... BCL2L1(598)

General description

Bcl-xL is a ~28 kDa member of Bcl-2 family of proteins and an important regulator of apoptosis. Bcl-xL forms heterodimers with BAX, BAK, and Bcl-2, and its overexpression in tumor cells confers resistance against chemotherapeutic drugs. Bcl-xL is phosphorylated on many sites including serine 62, a critical site for Bcl-xL response to microtubule-damaging drugs such as taxol and vinblastine. Phosphorylation of serine 62 (thought to be mediated by Jun N-terminal stress kinase (JNK) signaling) negatively regulates the anti-apoptotic function of Bcl-xL and controls the growth of neoplastic cells.

Immunogen

The antiserum was produced against a chemically synthesized phosphopeptide derived from the region of human Bcl-xL that contains serine 62. The sequence is conserved in mouse and rat.

Application

Anti-BCL-XL (pS62) Antibody detects level of BCL-XL (pS62) & has been published & validated for use in WB.
For Western blotting applications, we recommend using the antibody at a 1:1000 dilution.
Research Category
Apoptosis & Cancer
Research Sub Category
BCL2 & Inhibition

Biochem/physiol Actions

Human Bcl-xL. Mouse and rat Bcl-xL (100% conserved) have not been tested, but are expected to react.

Physical form

Dulbecco′s phosphate buffered saline (without Mg2+ and Ca2+), pH 7.3 (+/- 0.1), 50% glycerol, with 1.0 mg/mL BSA (IgG, protease free) as a carrier. 0.05% sodium azide.

Preparation Note

Store at -20°C. We recommend a brief centrifugation before opening to settle vial contents. Then, apportion into working aliquots and store at -20°C. For shipment or short-term storage (up to one week), 2-8°C is sufficient.

Analysis Note

Control
SK-BR-3 cells treated with vinblastine.
Purified from rabbit serum by sequential epitope-specific chromatography. The antibody has been negatively preadsorbed using a non-phosphopeptide corresponding to the site of phosphorylation to remove antibody that is reactive with non-phosphorylated Bcl-xL. The final product is generated by affinity chromatography using a Bcl-xL-derived peptide that is phosphorylated at serine 62.

Other Notes

Replaces: 04-432

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

WGK 2



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library