Skip to Content
Merck
All Photos(1)

Key Documents

AB3403-I

Sigma-Aldrich

Anti-Beta Galactosidase Antibody, E. coli

from chicken, purified by affinity chromatography

Synonym(s):

Beta-galactosidase, Acid beta-galactosidase, Lactase, Elastin receptor 1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€475.00

€475.00


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
100 μL
€475.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€475.00


Estimated to ship on29 May 2025


Request a Bulk Order

biological source

chicken

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

bacteria

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

Escherichia coli K12 ... Lacz(945006)

General description

β Galactosidase, also known as Beta-galactosidase, Beta-gal, LacZ, or Lactase, and encoded by the gene lacZ, is the enzyme in bacteria that hydrolyzes galactosides (sugars chains containing galactose) in to monosaccharides. β Galactosidase is typically an extracellularly produced enzyme that hydrolyzes the beta-glycosidic bond between galactose and another organic moiety. In E. coli, the β Galactosidase gene is present as part of the inducible lac operon system which is activated in the presence of lactose when glucose level is low. Today, bacterial beta galactosidase is used in molecular biology as a reporter marker to monitor gene expression, and because the enzyme has two functional domains, it forms the basis for α-complementation which is the methodology behind for the blue/white screening of recombinant clones. The presence or absence of an active β-galactosidase may be detected by X-gal, which produces a characteristic blue dye when cleaved by β-galactosidase, thereby providing an easy means of distinguishing the presence or absence of cloned product in a plasmid.

Immunogen

Recombinant protein corresponding to E. Coli Beta Galactosidase.

Application

Anti-Beta Galactosidase Antibody, E. coli is a highly specific chicken polyclonal antibody & that targets β-Galactosidase & has been tested in western blotting.

Quality

Evaluated by Western Blotting in TOP10/PCU19 E. Coli lysate.

Western Blotting Analysis: A 1:500 dilution of this antibody detected Beta Galactosidase in 10 µg TOP10/PCU19 E. Coli lysate.

Target description

~120 kDa observed

Linkage

Replaces: AB3403

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service