Skip to Content
Merck
All Photos(1)

Key Documents

AB3270

Sigma-Aldrich

Anti-K+-Cl- Cotransporter 4 Antibody

Chemicon®, from rabbit

Synonym(s):

KCC4

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
€335.00

€335.00


Estimated to ship on13 May 2025


Request a Bulk Order

Select a Size

Change View
50 μG
€335.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€335.00


Estimated to ship on13 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... SLC12A7(10723)

Specificity

Recognizes K+ Cl- Cotransporter-4 (KCC4). The immunogen shows no significant sequence homology with other KCCs or CCC.

SPECIES REACTIVITIES: The immunogen sequence is 81% homologous with human KCC4. Reactivity with other species has not been confirmed.

Immunogen

A 16 amino acid peptide within the cytoplasmic, N-terminus of mouse KCC4.

Application

Anti-K+-Cl-Cotransporter 4 Antibody is an antibody against K+-Cl-Cotransporter 4 for use in ELISA & WB.
Western blot: 1-10 μg/mL using ECL.

Approximate size on reduced westerns is 120-140kDa; membrane preparations and proteinase inhibitors are strongly recommended. See protocol under Technical Service as www.chemicon.com.



ELISA: 1:10,000-1:100,000 using 50-100 ng control peptide per well.

Optimal working dilutions must be determined by the end user.

Physical form

Affinity purified immunoglobulin. Liquid in 100 mM Tris, pH 7.5, containing 0.2% BSA and 0.05% sodium azide.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service