Skip to Content
Merck

AB3232

Anti-Low Density Lipoprotein Antibody, HOCL oxidized

serum, Chemicon®

Synonym(s):

LDL

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Clone:
polyclonal
Species reactivity:
human
Application:
ELISA, IHC
Citations:
1
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable, immunohistochemistry: suitable

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... LDLR(3949)

Immunogen

Epitope: HOCL oxidized
Hypochlorite-modified LDL.

Application

Immunohistochemistry: 1:50-1:400

ELISA: 1:400-1:4,000

Optimal working dilutions must be determined by the end user.
Research Category
Metabolism
Research Sub Category
Lipid Metabolism & Weight Regulation
This Anti-Low Density Lipoprotein Antibody, HOCL oxidized is validated for use in ELISA, IH for the detection of Low Density Lipoprotein.

Biochem/physiol Actions

Reacts strongly and very specifically with HOCL-LDL. By time resolved fluorescence there was no detectable cross reactivity to unmodified LDL has been observed. However weak cross reactivity with MDA-LDL (20%), HNE-LDL (5%) and minimally modified LDL (5%) was observed. By Western blot no detectable cross reactivity to unmodified LDL was observed. Weak cross reactivity (<10%) was detected with MDA-LDL, HNE-LDL and 24 hour Cu2+ oxidized LDL. By ELISA the antibody bound to early and late stages of HOCL-LDL but only weakly (<5%) to several other modifications including MDA-LDL, Cu2-ox-LDL, MDA-HSA and native LDL.

Physical form

Rabbit serum.

Preparation Note

Maintain lyophilized material at -20°C for up to 6 months After reconstitution maintain at -20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library