Skip to Content
Merck
All Photos(1)

Key Documents

AB3184

Sigma-Aldrich

Anti-MEK5 Antibody

Chemicon®, from rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
€310.00

€310.00


Estimated to ship on27 April 2025


Request a Bulk Order

Select a Size

Change View
50 μG
€310.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€310.00


Estimated to ship on27 April 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

Xenopus, canine, rabbit, sheep, monkey, human, mouse, rat

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... MAP2K5(5607)
mouse ... Map2K5(23938)

Specificity

MEK 5 is also known as MAP Kinase Kinase 5, or ERK Kinase 5. AB3184 detects a 49 kDa protein, corresponding to the expeted molecular mass of Mek5, in rat brain, rat liver, rat kidney, rat heart, mouse brain, and mouse testis tissue extracts as well as in cell lysates prepared from the rat pheochromacytoma (PC12) and the human HeLa cell lines. No cross-reactivity with MEK 1, 2, 3, 4 or 6 has been observed with this antibody.

Immunogen

Human MEK 5 synthetic peptide (aa. 59-74; Genbank Accession # U71088), conjugated to KLH; identical to rat MEK5 sequence over these residues.

Application

Anti-MEK5 Antibody is an antibody against MEK5 for use in ELISA, IH, IP & WB.
Research Category
Signaling
Research Sub Category
MAP Kinases
Western Blot: 1 μg/mL

Immunohistochemistry: 1:1,000

Immunoprecipitation

EIA

Optimal working dilutions must be determined by end user.

Physical form

Affinity purified immunoglobulin. Affinity purified immunoglobulin

Liquid containing PBS, pH 7.2. No preservatives.

Storage and Stability

Maintain frozen at -20°C in undiluted aliquots for up to 12 months from date of receipt. Thawed product may be stored at 2-8°C for up to 4 weeks. Avoid repeated freeze/thaw cycles.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pamela A Lochhead et al.
Cell cycle (Georgetown, Tex.), 15(4), 506-518 (2016-03-10)
ERK5, encoded by MAPK7, has been proposed to play a role in cell proliferation, thus attracting interest as a cancer therapeutic target. While oncogenic RAS or BRAF cause sustained activation of the MEK1/2-ERK1/2 pathway, ERK5 is directly activated by MEK5.
Pamela A Lochhead et al.
Nature communications, 11(1), 1383-1383 (2020-03-15)
The dual protein kinase-transcription factor, ERK5, is an emerging drug target in cancer and inflammation, and small-molecule ERK5 kinase inhibitors have been developed. However, selective ERK5 kinase inhibitors fail to recapitulate ERK5 genetic ablation phenotypes, suggesting kinase-independent functions for ERK5.
Jie Jiang et al.
Cell chemical biology, 27(12), 1553-1560 (2020-09-12)
MKK4/7 are kinases that phosphorylate JNKs and regulate the MAPK signaling pathway. Their overexpression has been associated with tumorigenesis and aggressiveness in cancers such as breast, prostate, non-small cell lung, and pediatric leukemia, making them a potential target for inhibitor
Fahima Munmun et al.
Journal of pineal research, 73(2), e12814-e12814 (2022-06-09)
Melatonin, the primary hormone involved in circadian entrainment, plays a significant role in bone physiology. This study aimed to assess the role of MEK1/2 and MEK5 in melatonin-mediated actions in mouse and human mesenchymal stem cells (MSCs) and on bone
Ravi K Komaravolu et al.
Cardiovascular research, 105(1), 86-95 (2014-11-13)
The MEK5/Erk5 pathway mediates beneficial effects of laminar flow, a major physiological factor preventing vascular dysfunction. Forced Erk5 activation induces a protective phenotype in endothelial cell (EC) that is associated with a dramatically decreased migration capacity of those cells. Transcriptional

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service