Skip to Content
Merck
All Photos(2)

Key Documents

AB2991

Sigma-Aldrich

Anti-p32 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

C1q globular domain-binding protein, GC1q-R protein, Glycoprotein gC1qBP, Hyaluronan-binding protein 1, Mitochondrial matrix protein p32, complement component 1, q subcomponent binding protein, splicing factor SF2-associated protein

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€385.00

€385.00


Estimated to ship on17 May 2025



Select a Size

Change View
100 μG
€385.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€385.00


Estimated to ship on17 May 2025


biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, rat, human

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... C1QBP(708)

General description

The p32 protein is a doughnut-shaped trimer that is primarily localized in the mitochondria. It has also been reported to be present at the cell surface and in the nucleus. The role of p32 in mammalian cells is unclear. The yeast p32 homologue has been reported to regulate oxidative phosphorylation, and a link to autophagy has been proposed. Elevated expression of p32 in cancer has also been noted. Additionally, p32 is primarily localized in hypoxic/nutrient-deprived regions within tumors and that, in addition to tumor cells, a tumor-associated macrophage/myeloid cell subpopulation closely linked to tumor lymphatics expresses high levels of p32. The expression of p32 at the cell surface, where it can be targeted with p32-binding antibodies and peptides, adds a second level of tumor specificity to p32 expression. The unique expression pattern of p32 in tumor cells, tumor lymphatics, and tumor-associated macrophages/myeloid cells makes p32 a potential target for the diagnosis and therapy of cancer.

Specificity

This antibody recognizes p32.

Immunogen

Cocktail of peptides corresponding to the N-terminus of human and mouse p32.
Epitope: Unknown

Application

Immunocytochemistry Analysis: 1:500 dilution from a previous lot detected p32 in HeLa and A431 cells.
Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional
This Anti-p32 Antibody is validated for use in WB, IC for the detection of p32.

Quality

Evaluated by Western Blot in A431 cell lysate.

Western Blot Analysis: 0.5 µg/ml of this antibody detected p32 on 10 µg of A431 cell lysate.

Target description

~ 32 kDa

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4, 150 mM NaCl) with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
A431 cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eric Savier et al.
Pharmaceutics, 13(10) (2021-10-24)
The interfering peptides that block protein-protein interactions have been receiving increasing attention as potential therapeutic tools. We measured the internalization and biological effect of four bi-functional tumor-penetrating and interfering peptides into primary hepatocytes isolated from three non-malignant and 11 hepatocellular
Maia Meparishvili et al.
Frontiers in behavioral neuroscience, 9, 319-319 (2015-12-05)
The intermediate and medial mesopallium (IMM) of the domestic chick forebrain has previously been shown to be a memory system for visual imprinting. Learning-related changes occur in certain plasma membrane and mitochondrial proteins in the IMM. Two-dimensional gel electrophoresis/mass spectrometry
Srinivas R Viswanathan et al.
Nature genetics, 50(7), 937-943 (2018-06-30)
Functional redundancy shared by paralog genes may afford protection against genetic perturbations, but it can also result in genetic vulnerabilities due to mutual interdependency1-5. Here, we surveyed genome-scale short hairpin RNA and CRISPR screening data on hundreds of cancer cell
Eduard Hofsetz et al.
Molecular & cellular proteomics : MCP, 19(8), 1330-1345 (2020-05-30)
The mammalian mitochondrial proteome consists of more than 1100 annotated proteins and their proteostasis is regulated by only a few ATP-dependent protease complexes. Technical advances in protein mass spectrometry allowed for detailed description of the mitoproteome from different species and
Karl Grenier et al.
The Journal of biological chemistry, 289(43), 29519-29530 (2014-09-14)
Parkinson disease (PD) is a complex neurodegenerative disease characterized by the loss of dopaminergic neurons in the substantia nigra. Multiple genes have been associated with PD, including Parkin and PINK1. Recent studies have established that the Parkin and PINK1 proteins

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service