Skip to Content
Merck
All Photos(2)

Documents

AB2267

Sigma-Aldrich

Anti-LCN2 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

lipocalin 2, migration-stimulating factor inhibitor, siderocalin, Oncogene 24p3, 25 kDa alpha-2-microglobulin-related subunit of MMP-9, lipocalin 2 (oncogene 24p3), neutrophil gelatinase-associated lipocalin, Lipocalin-2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

species reactivity (predicted by homology)

rat (based on 100% sequence homology), bovine (based on 100% sequence homology), mouse (based on 100% sequence homology)

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... LCN2(3934)

General description

LCN2 (also known as Neurophil Gelatinase-Associated Lipocalin or NGAL) belongs to a superfamily of lipocalins, a collection of small, diversely functional, extracellular proteins. NGAL is secreted from neutrophil granules in three different forms; a monomer of 25 kDa, a disulfide-linked homodimer at 46 kDa, and a disulfide-linked heterodimer with MMP-9 at 135-kDa. It is widely known to induce development of kidney epithelia, to bind and traffic iron, and to act as a molecular signal under inflammatory conditions, such as Crohn’s Disease. Multiple carcinoma studies have also shown elevated expression levels of LCN2 in several different human tumors affecting such organs as: colon, lung, breast, ovaries and pancreas. Additionally, there has been great interest in this protein as a possible marker for: onset of Acute Kidney Injury following cardiac surgery, the progression of Chronic Kidney Disease (CKD), and the survival chances of patients with Chronic Heart Failure (CHF).

Specificity

This antibody recognizes LCN2.

Immunogen

Epitope: Mature chain
KLH-conjugated linear peptide corresponding to the mature chain of human LCN2.

Application

Anti-LCN2 Antibody detects level of LCN2 & has been published & validated for use in WB, IH.
Immunohistochemistry Analysis: A 1:200 dilution of Anti-LCN2 antibody from a previous lot detected LCN2 in human spleen tissue.
Research Category
Neuroscience
Research Sub Category
Neurodegenerative Diseases

Quality

Evaluated by Western Blot in human kidney tissue lysate.

Western Blot Analysis: 5 µg/mL of this antibody detected LCN2 in 10 µg of human kidney tissue lysate.

Target description

LCN2 band observed at 25 kDa Heavy concentration of tissue lysate and a 5 minute exposure time were used to generate the band of interest.

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4, 150 mM NaCl) with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Human kidney tissue lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Emanuele Schiavon et al.
PloS one, 13(8), e0201022-e0201022 (2018-08-15)
Hyperbilirubinemia (jaundice) is caused by raised levels of unconjugated bilirubin in the blood. When severe, susceptible brain regions including the cerebellum and auditory brainstem are damaged causing neurological sequelae such as ataxia, hearing loss and kernicterus. The mechanism(s) by which
Mohammad A Alzubi et al.
Breast cancer research : BCR, 21(1), 36-36 (2019-03-08)
The seed and soil hypothesis was proposed over a century ago to describe why cancer cells (seeds) grow in certain organs (soil). Since then, the genetic properties that define the cancer cells have been heavily investigated; however, genomic mediators within
Reactive astrocytes secrete lcn2 to promote neuron death.
Bi, F; Huang, C; Tong, J; Qiu, G; Huang, B; Wu, Q; Li, F; Xu, Z; Bowser, R; Xia, XG; Zhou, H
Proceedings of the National Academy of Sciences of the USA null
Jingu Lee et al.
Glia, 70(5), 975-988 (2022-02-03)
Cerebral microinfarct increases the risk of dementia. But how microscopic cerebrovascular disruption affects the brain tissue in cellular-level are mostly unknown. Herein, with a longitudinal intravital imaging, we serially visualized in vivo dynamic cellular-level changes in astrocyte, pericyte and neuron
Zixian Huang et al.
Cell death & disease, 10(12), 936-936 (2019-12-11)
Chemoresistance is a major cause of cancer progression and the mortality of cancer patients. Developing a safe strategy for enhancing chemosensitivity is a challenge for biomedical science. Recent studies have suggested that vitamin D supplementation may decrease the risk of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service