Skip to Content
Merck
All Photos(1)

Key Documents

AB1966

Sigma-Aldrich

Anti-N-WASP Antibody, phospho-specific [Tyr256]

Chemicon®, from rabbit

Synonym(s):

Neural Wiskott-Aldrich Syndrome Protein

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€385.00

€385.00


Estimated to ship on30 May 2025


Request a Bulk Order

Select a Size

Change View
100 μL
€385.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€385.00


Estimated to ship on30 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... WASL(8976)

General description

Members of the Wiskott-Aldrich sydrome protein (WASP) family regulate the formation of actin-based cell structures in many cell types. These proteins contain C-terminal actin-binding domains that can stimulate actin polymerization. WASP is expressed primarily in hematopoietic cells, while its homolog N-WASP is widely expressed. These proteins have 48% identity in human with higher homology in the functional regions of these proteins. Phosphorylation at serine and tyrosine residues regulates the activity of both proteins. WASP is tyrosine phosphorylated at tyrosine 291 after antigen receptor activation in B-cells and collagen stimulation of platelets. Phosphorylation of the analogous site in N-WASP (Tyr256) stimulates its activity, reduces nuclear N-WASP, and is required for neurite extension.

Specificity

The antibody detects a 65 kDa protein corresponding to the molecular weight of phosphorylated N-WASP on SDS-PAGE Western blots of A431 cells treated with pervanadate. A similar band is observed in pervanadate treated HeLa and endothelial cells. Weak bands are also observed at higher molecular weights after pervanadate treatment. These bands may be due to low cross-reactivity with phosphotyrosine.

Immunogen

Epitope: phosphotyrosine 256
Phospho-N-WASP (Tyr256) synthetic peptide (coupled to BSA) corresponding to amino acid residues around tyrosine 256 of human N-WASP. The human WASP sequence has a two amino acid difference in the same region surrounding tyrosine 291.

Application

Anti-N-WASP Antibody, phospho-specific [Tyr256] detects level of N-WASP & has been published & validated for use in ELISA & WB.
Research Category
Signaling
Research Sub Category
Cytoskeletal Signaling
Western blot: 1:10,000 ELISA: 1:20,000

Physical form

This antibody was cross-adsorbed to phosphotyrosine then affinity purified using phospho-N-WASP (Tyr256) peptide (without carrier). Antibody is supplied in 100 μl phosphate-buffered saline, 50% glycerol, 1 mg/ml BSA, and 0.05% sodium azide.

Storage and Stability

Maintain at 20°C for up to one year from date of receipt. Do not aliquot.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yinna Wang et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 35(9), e21811-e21811 (2021-08-10)
Actin cytoskeletal reorganization plays an important role in regulating smooth muscle contraction, which is essential for the modulation of various physiological functions including airway tone. The adapter protein Abi1 (Abelson interactor 1) participates in the control of smooth muscle contraction.
Yinna Wang et al.
American journal of respiratory cell and molecular biology, 62(5), 645-656 (2020-01-09)
It has been reported that actin polymerization is regulated by protein tyrosine phosphorylation in smooth muscle on contractile stimulation. The role of protein serine/threonine phosphorylation in modulating actin dynamics is underinvestigated. SLK (Ste20-like kinase) is a serine/threonine protein kinase that
Ruping Wang et al.
Scientific reports, 10(1), 10667-10667 (2020-07-02)
Smooth muscle cell migration is essential for many diverse biological processes such as pulmonary/cardiovascular development and homeostasis. Abi1 (Abelson interactor 1) is an adapter protein that has been implicated in nonmuscle cell migration. However, the role and mechanism of Abi1
Xuejiao Sun et al.
Biochemistry and cell biology = Biochimie et biologie cellulaire, 100(6), 458-472 (2022-11-08)
FYCO1, an autophagy adaptor, plays an essential role in the trafficking toward the plus-end of microtubules and the fusion of autophagosomes. Autophagic dysfunction is involved in numerous disease states, including cancers. Previous studies have implicated FYCO1 as one of the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service