Skip to Content
Merck
All Photos(1)

Key Documents

AB16955

Sigma-Aldrich

Anti-DR4 Antibody, CT

Chemicon®, from rabbit

Synonym(s):

TRAIL-R1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€327.00

€327.00


Estimated to ship on14 April 2025


Request a Bulk Order

Select a Size

Change View
100 μG
€327.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€327.00


Estimated to ship on14 April 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

Specificity

Apoptosis, or programmed cell death, occurs during normal cellular diffferentiation and development of multicellular organisms. Apoptosis is induced by certain cytokines including TNF and Fas ligand of the TNF family through their death domain containing receptors, TNFR1 and Fas. A novel cell death receptor was recently identified by several groups independently and designated DR4 (for death receptor 4) (Pan et al. 1997). The ligand for this novel cell death receptor has been identified and termed TRAIL, which is a new member in TRF family. Therefore, DR4 is also called TRAIL receptor-1 (TRAIL-R1) (Schneider et al. 1997). DR4 is expresssed in most of human tissues including spleen, peripheral blood leukocytes, small intestine and thymus. Like TNFR1, Fas and DR3, DR4 mediates TRAIL induced apoptosis and NF-kB activation in many tissues and cells.

Immunogen

Epitope: C-terminus
Polyclonal antibody raised against a peptide corresponding to amino acids 427-445 at C-terminus of human DR4 mature protein.

Application

Detect DR4 using this Anti-DR4 Antibody, C-terminus validated for use in WB.
Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional
Western blot: 1:500-1:1000

Whole cell lysate from HeLa cells can be used as a positive control and a 57 kDa band should be detected.

Optimal working dilutions must be determined by end user.

Physical form

Affinity purified immunoglobulin. Liquid in PBS containing 0.02% sodium azide.
Format: Purified

Storage and Stability

Maintain refrigerated at 2-8°C in undiluted aliquots for up to 12 months.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

V Pavet et al.
Cell death & disease, 5, e1043-e1043 (2014-02-01)
Tumor necrosis factor-related apoptosis-inducing ligand (TRAIL/TNFSF10/Apo2L) holds promise for cancer therapy as it induces apoptosis in a large variety of cancer cells while exerting negligible toxicity in normal ones. However, TRAIL can also induce proliferative and migratory signaling in cancer
Andrei Alexandru Constantinescu et al.
Methods in molecular biology (Clifton, N.J.), 1557, 19-31 (2017-01-13)
Analysis of CD95/Fas complexes by immunoprecipitation has long relied on the monoclonal antibody APO1 or tagged recombinant Fas ligand. Immunoprecipitation is an elegant and efficient procedure to investigate endogenous protein interactions or complexes. Provided that the targeted complex is soluble
Anna Willms et al.
PloS one, 14(4), e0214847-e0214847 (2019-04-05)
Due to their ability to preferentially induce cell death in tumor cells, while sparing healthy cells, TNF-related apoptosis-inducing ligand (TRAIL) and agonistic anti-TRAIL-R1 or anti-TRAIL-R2-specific antibodies are under clinical investigations for cancer-treatment. However, TRAIL-Rs may also induce signaling pathways, which
Marie-Laure Plissonnier et al.
Oncotarget, 8(64), 107744-107762 (2018-01-04)
Known activators of the Peroxisome Proliferator-Activated Receptor γ (PPARγ), thiazolidinediones (TZD) induce apoptosis in a variety of cancer cells through dependent and/or independent mechanisms of the receptor. We tested a panel of TZD (Rosiglitazone, Pioglitazone, Ciglitazone) to shed light on
Florent Dufour et al.
Oncotarget, 8(6), 9974-9985 (2017-01-01)
TRAIL induces selective tumor cell death through TRAIL-R1 and TRAIL-R2. Despite the fact that these receptors share high structural homologies, induction of apoptosis upon ER stress, cell autonomous motility and invasion have solely been described to occur through TRAIL-R2. Using

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service