Skip to Content
Merck
All Photos(1)

Key Documents

AB16926

Sigma-Aldrich

Anti-DFF40 Antibody

Chemicon®, from rabbit

Synonym(s):

DNA Fragmentation Factor, 40 kDa Subunit, Caspase-activated DNase, CAD, CPAN

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€544.00

€544.00


Estimated to ship on24 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
€544.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€544.00


Estimated to ship on24 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, mouse, rat

manufacturer/tradename

Chemicon®

technique(s)

western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... DFFB(1677)

Specificity

Apoptosis is related to many diseases and induced by a family of cell death receptors and their ligands. Cell death signals are transduced by death domain containing adapter molecules and members of the caspase family of proteases. These death signals finally cause the degradation of chromosomal DNA by activated DNase. A mouse DNase that causes DNA fragmentation was identified recently and designated CAD (for caspase activated deoxyribonuclease) (Enari et al. 1998; Sakahira et al. 1998). The human homologue of mouse CAD was more recently identified by three groups independently and termed CPAN and DFF40 and human CAD, respectively (Halenbeck et al. 1998; Liu et al. 1998; Mukae et al. 1998). DFF45/ICAD is the inhibitory protein of DFF40/CAD (Enari et al. 1998; Sakahira et al. 1998; Liu et al. 1997) and forms complex with DFF40/CAD. Upon cleavage of DFF45/ICAD by activated caspase, DFF40/CAD is released and activated and eventually cuases the degradation of DNA in the nuclei. Activation of DFF40/CAD, which causes DNA degradation, is the hallmark of apoptotic cell death.

Immunogen

A peptide corresponding to amino acids 147-164 of mouse CAD (Enari et al. 1998). The sequence differs from human DFF40 by two amino acids (Halenbeck et al. 1998; Liu et al. 1998).

Application

Detect DFF40 using this Anti-DFF40 Antibody validated for use in WB.
Western blot: 1:500-1:1,000

K562 or Jurkat whole cell lysate can be used as a positive control and a 40 kDa band should be detected.

Optimal working dilutions must be determined by end user.

Physical form

Format: Purified

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aiying Du et al.
Cell discovery, 1, 15002-15002 (2015-01-01)
In addition to terminating neurotransmission by hydrolyzing acetylcholine, synaptic acetylcholinesterase (AChES) has been found to have a pro-apoptotic role. However, the underlying mechanism has rarely been investigated. Here, we report a nuclear translocation-dependent role for AChES as an apoptotic deoxyribonuclease
María Sánchez-Osuna et al.
The Journal of biological chemistry, 289(27), 18752-18769 (2014-05-20)
Caspase-dependent apoptosis is a controlled type of cell death characterized by oligonucleosomal DNA breakdown and major nuclear morphological alterations. Other kinds of cell death do not share these highly distinctive traits because caspase-activated DNase (DFF40/CAD) remains inactive. Here, we report
Charles W Morgan et al.
Proceedings of the National Academy of Sciences of the United States of America, 112(27), 8344-8349 (2015-06-25)
Cellular demolition during apoptosis is completed by executioner caspases, that selectively cleave more than 1,500 proteins but whose individual roles are challenging to assess. Here, we used an optimized site-specific and inducible protease to examine the role of a classic
B Gole et al.
Oncogene, 34(26), 3391-3401 (2014-08-19)
MLL (myeloid/lymphoid or mixed-lineage leukemia) rearrangements are frequent in therapy-related and childhood acute leukemia, and are associated with poor prognosis. The majority of the rearrangements fall within a 7.3-kb MLL breakpoint cluster region (MLLbcr), particularly in a 0.4-kb hotspot at
Victoria Iglesias-Guimarais et al.
The Journal of biological chemistry, 288(13), 9200-9215 (2013-02-23)
Apoptotic nuclear morphology and oligonucleosomal double-strand DNA fragments (also known as DNA ladder) are considered the hallmarks of apoptotic cell death. From a classic point of view, these two processes occur concomitantly. Once activated, DNA fragmentation factor, 40-kDa subunit (DFF40)/caspase-activated

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service