Skip to Content
Merck
All Photos(1)

Key Documents

AB1567

Sigma-Aldrich

Anti-Luteinizing Hormone Releasing Hormone Antibody

serum, Chemicon®

Synonym(s):

LHRH, GNRH

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€356.00

€356.00


Estimated to ship on26 May 2025


Request a Bulk Order

Select a Size

Change View
100 μL
€356.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€356.00


Estimated to ship on26 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

sheep, mouse, chicken, fish, human, rat, rabbit

manufacturer/tradename

Chemicon®

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable (paraffin)

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... GNRH1(2796)

Specificity

Luteinizing Hormone Releasing Hormone (LHRH)

Immunogen

LHRH (Sigma) conjugated to thyroglobulin.

Application

Anti-Luteinizing Hormone Releasing Hormone Antibody detects level of Luteinizing Hormone Releasing Hormone & has been published & validated for use in IC, IH(P).
Immunohistochemistry: 1:5,000-1:10,000 (18-24 hour incubation) on formalin, Bouin′s or Zamboni′s fixed tissue. Will react on paraffin embedded tissue.

Immunocytochemistry: 1:10,000 on fetal rat neurons.

Optimal working dilutions must be determined by the end user.
Research Category
Neuroscience
Research Sub Category
CNS Control of Metabolism

Hormones & Receptors

Linkage

Replaces: AB1756

Physical form

Neat rabbit antisera. Liquid

Storage and Stability

Store at -20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tipaporn Kumkoon et al.
Toxins, 15(4) (2023-04-27)
Breast cancer is one of the most common cancers in women worldwide. Conventional cancer chemotherapy always has adverse side effects on the patient's healthy tissues. Consequently, combining pore-forming toxins with cell-targeting peptides (CTPs) is a promising anticancer strategy for selectively
Martin Meschkat et al.
Nature communications, 13(1), 1163-1163 (2022-03-06)
Myelin, the electrically insulating sheath on axons, undergoes dynamic changes over time. However, it is composed of proteins with long lifetimes. This raises the question how such a stable structure is renewed. Here, we study the integrity of myelinated tracts
Xiaoxuan Ren et al.
Neural regeneration research, 7(31), 2413-2423 (2012-11-05)
In mammals, gonadal function is controlled by the activity of hypothalamic gonadotropin-releasing hormone neurons, which control the secretion of adenohypophyseal and gonadal hormones. However, there are a number of unanswered questions in relation to gonadal function. It is currently unknown

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service