Skip to Content
Merck
All Photos(1)

Documents

AB15520

Sigma-Aldrich

Anti-Neuronal pentraxin 2 Antibody

Chemicon®, from rabbit

Synonym(s):

NPX2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

mouse ... Morc3(338467)

General description

Neuronal Pentraxins: A new family of putative integral membrane pentraxins or Neuronal pentraxins identified through interaction with a presynaptic snake venom toxin taipoxin. NPX1, NPX2 and NPXR (Neuronal Pentraxin Receptor), the three neuronal pentraxins represent a novel neuronal uptake pathway that may function during synapse formation and remodeling. The N-terminal half of neuronal pentraxins are 20-30% identical to previously identified pentraxins (CRP and SAP), the three neuronal pentraxins are 50% identical to each other and are significantly larger than the classical pentraxins (>50 versus 30kDa), suggesting that they may have additional novel functions. NPX2 (Neuronal Pentraxin 2), 429aa in mouse, 432 in rat and 431 in human (chr 7q21), mainly expressed in brain, pancrease, liver and testis. It is likely to play role in the modification of cellular properties that underlie long-term plasticity. NPX1, NPX2 has potential N-linked glycosylation sites.

Specificity

Other species have not yet been tested. The immunogen sequence is 100% conserved in rat, and 94% in human NPX2 and 82% in guinea pig apexin precursor P50.

Immunogen

Synthetic peptide near the N-terminus of mouse NPX2

Application

Detect Neuronal pentraxin 2 using this Anti-Neuronal pentraxin 2 Antibody validated for use in ELISA & WB.
Research Category
Neuroscience
Research Sub Category
Synapse & Synaptic Biology
Western blot: 1-10 µg/mL using ECL.
Immunohistochemistry: Not tested. It is recommended that the antibody be tried at 2-20 µg/mL
ELISA: 1:10,000-1:100,000 using 50-100 ng of control peptide/well.
Optimal working dilutions must be determined by end user.

Physical form

Format: Purified
ImmunoAffinity Purified
Liquid in 100 mM Tris, pH 7.5 and 0.1% BSA.

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 6 months after date of receipt. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service