Skip to Content
Merck
All Photos(1)

Key Documents

AB1537

Sigma-Aldrich

Anti-Dopamine β Hydroxylase Antibody, CT

Chemicon®, from sheep

Synonym(s):

DBH

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€572.00

€572.00


Estimated to ship on01 June 2025

A recombinant, preservative-free antibody is available for your target. Try ZRB1328

Request a Bulk Order

Select a Size

Change View
100 μL
€572.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€572.00


Estimated to ship on01 June 2025

A recombinant, preservative-free antibody is available for your target. Try ZRB1328

Request a Bulk Order

biological source

sheep

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

monkey, human

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... DBH(1621)

Immunogen

Synthetic peptide, CPTSQGRSPAGPTVVSI-amide, from C-terminal region of human (DbetaH).

Application

Immunoblotting: 1:250-1:1,000 on human, monkey and bovine adrenal medulla.

Immunohistochemistry: 1:250-1:1,000, staining was evaluated in the locus coeruleus of formaldehyde-fixed, cryopreserved human and monkey brain. Showed strong staining on both human and monkey.

Optimal working dilutions must be determined by the end user.
This Anti-Dopamine β Hydroxylase Antibody, C-terminus is validated for use in WB, IH for the detection of Dopamine β Hydroxylase.

Physical form

Affinity purified immunoglobulin. Liquid in 10 mM HEPES, pH 7.5, 150 mM NaCl, 100 μg/mL BSA and 50% glycerol.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Isabel Pérez-Santos et al.
Cerebral cortex (New York, N.Y. : 1991), 31(9), 4115-4139 (2021-05-19)
Noradrenaline (NA) in the thalamus has important roles in physiological, pharmacological, and pathological neuromodulation. In this work, a complete characterization of NA axons and Alpha adrenoceptors distributions is provided. NA axons, revealed by immunohistochemistry against the synthesizing enzyme and the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service