Skip to Content
Merck
All Photos(1)

Key Documents

AB1518

Sigma-Aldrich

Anti-Neurofibrillary Tangles Antibody

serum, Chemicon®

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€586.00

€586.00


Estimated to ship on20 May 2025


Request a Bulk Order

Select a Size

Change View
100 μL
€586.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€586.00


Estimated to ship on20 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

shipped in

dry ice

target post-translational modification

unmodified

Specificity

Specifically stains neurofibrillary tangles and degenerating plaque neurites in cases of Alzheimer′s disease, Down′s Syndrome and normal aged individuals.

Immunogen

Neurofibrillary tangles which were extracted in boiling SDS and purified by sucrose gradient centrifugation.

Application

Anti-Neurofibrillary Tangles Antibody is an antibody against Neurofibrillary Tangles for use in ELISA, WB, IH, IH(P).
Immunohistochemistry on paraffin, vibratome and frozen sections: ≥1:200, ≥1:2,000, respectively.

Western blotting: ≥1:500.

ELISA using immunogen peptide: ≥1:4,000.

Optimal working dilutions must be determined by end user.
Research Category
Neuroscience
Research Sub Category
Neurodegenerative Diseases

Physical form

Rabbit antisera with 0.01% thimerosal.

Storage and Stability

Store at -20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reducing amyloid-related Alzheimer's disease pathogenesis by a small molecule targeting filamin A.
Wang, HY; Bakshi, K; Frankfurt, M; Stucky, A; Goberdhan, M; Shah, SM; Burns, LH
The Journal of Neuroscience null
Milena Penkowa et al.
Journal of neuroscience research, 77(1), 35-53 (2004-06-16)
Transgenic expression of interleukin-6 (IL-6) in the CNS under the control of the glial fibrillary acidic protein (GFAP) gene promoter (GFAP-IL6 mice) induces significant inflammation and neurodegeneration but also affords neuroprotection against acute traumatic brain injury. This neuroprotection is likely
Julio J Ramirez et al.
Behavioural brain research, 216(1), 332-340 (2010-08-24)
Entorhinal cortex neuropathology begins very early in Alzheimer's disease (AD), a disorder characterized by severe memory disruption. Indeed, loss of entorhinal volume is predictive of AD and two of the hallmark neuroanatomical markers of AD, amyloid plaques and neurofibrillary tangles
Milena Penkowa et al.
Journal of neuroscience research, 79(4), 522-534 (2004-12-23)
We examined metallothionein (MT)-induced neuroprotection during kainic acid (KA)-induced excitotoxicity by studying transgenic mice with MT-I overexpression (TgMT mice). KA induces epileptic seizures and hippocampal excitotoxicity, followed by inflammation and delayed brain damage. We show for the first time that
Leslie Vargas-Saturno et al.
The European journal of neuroscience, 48(12), 3514-3533 (2018-11-08)
Understanding endogenous neurogenesis and neuronal replacement to mature circuits is a topic of discussion as a therapeutic alternative under acute and chronic neurodegenerative disorders. Adaptive neurogenic response may result as a result of ischemia which could support long-term recovery of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service