Skip to Content
Merck
All Photos(2)

Key Documents

AB15097

Sigma-Aldrich

Anti-mGluR4 Antibody

from rabbit

Synonym(s):

glutamate metabotropic receptor 4, glutamate receptor, metabotropic 4

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
€385.00

€385.00


Estimated to ship on14 May 2025


Request a Bulk Order

Select a Size

Change View
50 μG
€385.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€385.00


Estimated to ship on14 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse, rat

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

mouse ... Grm4(268934)

General description

L-glutamate is the major excitatory neurotransmitter in the central nervous system and activates both ionotropic and metabotropic glutamate receptors. Glutamatergic neurotransmission is involved in most aspects of normal brain function and can be perturbed in many neuropathologic conditions. The metabotropic glutamate receptors are a family of G protein-coupled receptors that have been divided into 3 groups on the basis of sequence homology, putative signal transduction mechanisms, and pharmacologic properties. Group I includes GRM1 and GRM5 and these receptors have been shown to activate phospholipase C. Group II includes GRM2 and GRM3 while Group III includes GRM4, GRM6, GRM7 and GRM8. Group II and III receptors are linked to the inhibition of the cyclic AMP cascade but differ in their agonist selectivities.

Specificity

Cat. # AB15097 recognizes mGluR4 (GRM4).
Reactivity with other species has not been determined.

Immunogen

Recombinant Protein.

Application

Research Category
Neuroscience
Research Sub Category
Neurotransmitters & Receptors
This Anti-mGluR4 Antibody is validated for use in WB for the detection of mGluR4.

Quality

Western Blot:
Mouse Brain Lysate.

Target description

~101 kDa

Physical form

Antigen Affinity Purified
Format: Purified
Purified in 0.1M Tris-Glycine (pH7.4) 150mM NaCl with 0.05% NaN3.

Storage and Stability

Stable for 1 year at 2-8ºC from date of receipt.

Analysis Note

Control
Mouse Brain Lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Native presynaptic metabotropic glutamate receptor 4 (mGluR4) interacts with exocytosis proteins in rat cerebellum.
Ramos, C; Chardonnet, S; Marchand, CH; Decottignies, P; Ango, F; Daniel, H; Le Marechal, P
The Journal of Biological Chemistry null
Balaji Krishnan et al.
Neurobiology of learning and memory, 128, 65-79 (2016-01-10)
Long-term memory (LTM) of fear stores activity dependent modifications that include changes in amygdala signaling. Previously, we identified an enhanced probability of release of glutamate mediated signaling to be important in rat fear potentiated startle (FPS), a well-established translational behavioral
Syutaro Toya et al.
The European journal of neuroscience, 40(11), 3627-3634 (2014-09-16)
Early-life stress induces several neuropsychological disorders in adulthood, including depression. Such disorders may be induced by functional alteration of the glutamatergic system. However, their underlying mechanisms have not yet been fully clarified. Furthermore, the involvement of glucocorticoids, which are representative
Caesar M Hernandez et al.
eNeuro, 5(3) (2018-07-05)
Glutamate signaling is essential for the persistent neural activity in prefrontal cortex (PFC) that enables working memory. Metabotropic glutamate receptors (mGluRs) are a diverse class of proteins that modulate excitatory neurotransmission via both presynaptic regulation of extracellular glutamate levels and
Anna V Golubeva et al.
Physiological reports, 2(3), e00260-e00260 (2014-04-25)
Abstract Painful bladder syndrome/Interstitial cystitis (PBS/IC) is a chronic disorder characterized clinically by recurring episodes of pelvic pain and increased urination frequency, significantly impairing patients' quality of life. Despite this, there is an unmet medical need in terms of effective

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service