Skip to Content
Merck
All Photos(1)

Key Documents

AB135

Sigma-Aldrich

Anti-Tryptophan Antibody

serum, Chemicon®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702

biological source

rabbit

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

rat

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable

shipped in

dry ice

target post-translational modification

unmodified

Specificity

Tryptophan

The cross-reactivities were determined using an ELISA test by competition experiments with the following compounds:

Compound Cross-reactivity
Tryptophan-G-PL 1
5-Hydroxytryptophan-G-PL .01
5-Methoxytryptophan-G-PL .004
Tryptophan=G=PL (non reduced) .002
Tryptophan .0003
Tryptamine-G-PL .00009
5-Methoxytryptamine-G-PL .00002
5-Hydroxytryptamine-G-PL .000004

The antisera was also tested for specificity using the free-floating PAP technique on rat brain.

Abbreviations:
(G)Glutaraldehyde
(=)Non-reduced conjugate
(PL)Poly-lysine

Immunogen

Tryptophan-gluteraldehyde-Poly-lysine

Application

Anti-Tryptophan Antibody is an antibody against Tryptophan for use in IH.
Immunohistochemistry: 1:1,000-1:2,500 by PAP (see suggested protocol).

Optimal working dilutions must be determined by the end user.
Research Category
Neuroscience
Research Sub Category
Neurotransmitters & Receptors

Physical form

Rabbit antiserum. Liquid with 0.05% sodium azide.

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service