Skip to Content
Merck
All Photos(1)

Key Documents

AB1225

Sigma-Aldrich

Anti-Ovalbumin Antibody, Egg white

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€335.00

€335.00


Estimated to ship on20 May 2025


Request a Bulk Order

Select a Size

Change View
100 μL
€335.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43

€335.00


Estimated to ship on20 May 2025


Request a Bulk Order

antibody form

purified antibody

Quality Level

clone

polyclonal

species reactivity

rabbit, chicken

concentration

(Please refer to the Certificate of Analysis for the lot-specific concentration.)

technique(s)

ELISA: suitable
immunohistochemistry: suitable
western blot: suitable

UniProt accession no.

General description

Ovalbumin (Hen Egg White)

Specificity

Ovalbumin (Hen Egg White). Monospecific by IEP.

Application

Western blot: 1:500-1:2,000
Immunohistochemistry: 1:300-1:2,000
ELISA: 1:3,000-1:12,000 when assayed against 1 μg of Ovalbumin in a standard Indirect ELISA
Optimal working dilutions must be determined by end user.

Physical form

Immunoglobulin purified by delipidation, salt fractionation and ion-exchange chromatography. Liquid in 0.02 M Potassium Phosphate buffer, 0.15 M Sodium Chloride, pH 7.2, with 0.01% sodium azide.

Storage and Stability

Maintain frozen at -20°C in undiluted aliquots for up to six months after date of receipt. Avoid repeated freeze/thaw cycles.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service