Skip to Content
Merck
All Photos(3)

Key Documents

AB10535

Sigma-Aldrich

Anti-Neurogenin-3 (rabbit) Antibody

serum, from rabbit

Synonym(s):

Neurogenin 3, NGN3, Neurogenin3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

mouse ... Neurog3(11925)

General description

Neurogenins are a family of bHLH transcription factors involved in specifying neuronal differentiation. They are related to Drosophila atonal. The neurogenins (ngns) make up one of these atonal-related gene families. In neural crest cells, the atonal-related neurogenin family is particularly important for the sensory lineage (Neurogenins Are Essential for the Formation of dorsal root ganglia), while the achaete-scute homologue ash1 (Mash1) is important for aspects of autonomic neurogenesis (mash1 is essential for noradrenergic differentiation). Neurogenin-3 also promotes pancreatic development.

Specificity

This antibody recognizes Neurogenin-3 at the N-terminus.

Immunogen

Epitope: N-terminus
GST-tagged recombinant protein corresponding to the N-terminus of mouse Neurogenin-3.

Application

Detect Neurogenin-3 (rabbit) using this Anti-Neurogenin-3 (rabbit) Antibody validated for use in WB, IH.
Immunohistochemistry Analysis: A 1:500 dilution from a previous lot detected Neurogenin-3 in mouse embryo E12.5+ tissue.
Research Category
Neuroscience
Research Sub Category
Developmental Neuroscience

Quality

Evaluated by Western Blot in mNgn3 transfected and wildtype rabbit reticulocyte lysates.

Western Blot Analysis: A 1:1,000 dilution of this antibody detected Neurogenin-3 on 10 µg of mNgn3 transfected rabbit reticulocyte lysates.

Target description

32 kDa

Physical form

Unpurified
Unpurified rabbit polyclonal serum containing 0.05% sodium azide.

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
mNgn3 transfected and wildtype rabbit reticulocyte lysates

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service